Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39029188-39029433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3D
Entrez Gene ID
140564
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3D
Gene Aliases
A3D, APOBEC3DE, APOBEC3E, ARP6
UniGene
Hs.629775
Gene Chromosome Location
Chr.22: 39021113-39034328 on Build GRCh38
Transcripts
NM_152426.3 XM_005261337.3 XM_017028596.1 XM_017028643.1

Gene Symbol
APOBEC3F
Entrez Gene ID
200316
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3F
Gene Aliases
A3F, ARP8, BK150C2.4.MRNA, KA6
UniGene
Hs.659809
Gene Chromosome Location
Chr.22: 39020454-39055972 on Build GRCh38
Transcripts
NM_152426.3 XM_005261337.3 XM_017028596.1 XM_017028643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6232447 c.641A>T p.Y214F Chr.22: 39029398-39029398 on Build GRCh38
5057820 c.662T>C p.L221P Chr.22: 39029419-39029419 on Build GRCh38
5748261 c.673G>A p.G225S Chr.22: 39029430-39029430 on Build GRCh38
292627 c.652A>G p.K218E Chr.22: 39029409-39029409 on Build GRCh38
3800205 c.612G>A p.P204P Chr.22: 39029369-39029369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766169459 Chr.22: 39029318-39029318 on Build GRCh38
rs563139699 Chr.22: 39029288-39029288 on Build GRCh38
rs776510359 Chr.22: 39029328-39029329 on Build GRCh38
rs577361405 Chr.22: 39029295-39029295 on Build GRCh38
rs141550709 Chr.22: 39029289-39029289 on Build GRCh38
rs779214802 Chr.22: 39029369-39029369 on Build GRCh38
rs548520941 Chr.22: 39029274-39029274 on Build GRCh38
rs756881964 Chr.22: 39029261-39029261 on Build GRCh38
rs567021538 Chr.22: 39029287-39029287 on Build GRCh38
rs373725653 Chr.22: 39029393-39029393 on Build GRCh38
rs542273414 Chr.22: 39029337-39029337 on Build GRCh38
rs770840029 Chr.22: 39029238-39029238 on Build GRCh38
rs746588206 Chr.22: 39029221-39029221 on Build GRCh38
rs780647042 Chr.22: 39029380-39029380 on Build GRCh38
rs117814205 Chr.22: 39029323-39029323 on Build GRCh38
rs757469920 Chr.22: 39029368-39029368 on Build GRCh38
rs746174647 Chr.22: 39029378-39029378 on Build GRCh38
rs556558970 Chr.22: 39029322-39029322 on Build GRCh38
rs777893956 Chr.22: 39029352-39029352 on Build GRCh38
rs959800894 Chr.22: 39029263-39029263 on Build GRCh38
rs969772663 Chr.22: 39029235-39029235 on Build GRCh38
rs908725744 Chr.22: 39029279-39029279 on Build GRCh38
rs538236336 Chr.22: 39029300-39029300 on Build GRCh38
rs776492670 Chr.22: 39029266-39029266 on Build GRCh38
rs992247857 Chr.22: 39029267-39029267 on Build GRCh38
rs756172183 Chr.22: 39029346-39029346 on Build GRCh38
rs1050207290 Chr.22: 39029385-39029385 on Build GRCh38
rs371165008 Chr.22: 39029357-39029357 on Build GRCh38
rs752637566 Chr.22: 39029340-39029340 on Build GRCh38
rs772487858 Chr.22: 39029379-39029379 on Build GRCh38

View Full Product Details