Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103325411-103325652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD6
Entrez Gene ID
8323
Gene Name
frizzled class receptor 6
Gene Aliases
FZ-6, FZ6, HFZ6, NDNC10
UniGene
Hs.591863
Gene Chromosome Location
Chr.8: 103298433-103332866 on Build GRCh38
Transcripts
NM_001164615.1 NM_001164616.1 NM_001317796.1 NM_003506.3 NR_133921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6054221 c.1350C>G p.V450V Chr.8: 103325456-103325456 on Build GRCh38
3884265 c.1375A>G p.I459V Chr.8: 103325481-103325481 on Build GRCh38
6668169 c.1306G>A p.V436I Chr.8: 103325412-103325412 on Build GRCh38
3269726 c.1390C>G p.Q464E Chr.8: 103325496-103325496 on Build GRCh38
5727414 c.1308C>T p.V436V Chr.8: 103325414-103325414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963976545 Chr.8: 103325481-103325481 on Build GRCh38
rs1001103495 Chr.8: 103325545-103325545 on Build GRCh38
rs781683461 Chr.8: 103325573-103325573 on Build GRCh38
rs139148335 Chr.8: 103325486-103325486 on Build GRCh38
rs12707777 Chr.8: 103325620-103325620 on Build GRCh38
rs779693042 Chr.8: 103325498-103325498 on Build GRCh38
rs774874477 Chr.8: 103325528-103325528 on Build GRCh38
rs1018228524 Chr.8: 103325480-103325480 on Build GRCh38
rs568705981 Chr.8: 103325593-103325593 on Build GRCh38
rs531164438 Chr.8: 103325565-103325565 on Build GRCh38
rs757357454 Chr.8: 103325446-103325446 on Build GRCh38
rs746771854 Chr.8: 103325508-103325508 on Build GRCh38
rs747714107 Chr.8: 103325522-103325522 on Build GRCh38
rs551894404 Chr.8: 103325470-103325470 on Build GRCh38
rs780764882 Chr.8: 103325517-103325517 on Build GRCh38
rs764886770 Chr.8: 103325468-103325468 on Build GRCh38
rs368662779 Chr.8: 103325489-103325489 on Build GRCh38
rs368015730 Chr.8: 103325469-103325469 on Build GRCh38
rs754589676 Chr.8: 103325515-103325515 on Build GRCh38
rs371900641 Chr.8: 103325574-103325574 on Build GRCh38
rs746335327 Chr.8: 103325533-103325533 on Build GRCh38
rs769289936 Chr.8: 103325525-103325525 on Build GRCh38
rs1009721358 Chr.8: 103325613-103325613 on Build GRCh38
rs889121099 Chr.8: 103325539-103325539 on Build GRCh38
rs527378976 Chr.8: 103325454-103325454 on Build GRCh38

View Full Product Details