Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34794448-34794705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA4
Entrez Gene ID
2701
Gene Name
gap junction protein alpha 4
Gene Aliases
CX37
UniGene
Hs.296310
Gene Chromosome Location
Chr.1: 34792958-34795747 on Build GRCh38
Transcripts
NM_002060.2 XM_005270750.2 XM_017001043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
426147 c.247C>G p.L83V Chr.1: 34794460-34794460 on Build GRCh38
6039913 c.373C>T p.R125W Chr.1: 34794586-34794586 on Build GRCh38
7369475 c.341G>A p.R114Q Chr.1: 34794554-34794554 on Build GRCh38
2079902 c.469G>A p.A157T Chr.1: 34794682-34794682 on Build GRCh38
5762022 c.258C>T p.S86S Chr.1: 34794471-34794471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774646282 Chr.1: 34794515-34794515 on Build GRCh38
rs757382942 Chr.1: 34794643-34794643 on Build GRCh38
rs770022924 Chr.1: 34794478-34794478 on Build GRCh38
rs754112258 Chr.1: 34794622-34794622 on Build GRCh38
rs764909085 Chr.1: 34794533-34794533 on Build GRCh38
rs777931705 Chr.1: 34794484-34794484 on Build GRCh38
rs753372796 Chr.1: 34794531-34794531 on Build GRCh38
rs530865940 Chr.1: 34794560-34794560 on Build GRCh38
rs763601338 Chr.1: 34794527-34794527 on Build GRCh38
rs777775195 Chr.1: 34794586-34794586 on Build GRCh38
rs771173925 Chr.1: 34794507-34794507 on Build GRCh38
rs750230827 Chr.1: 34794536-34794536 on Build GRCh38
rs758289830 Chr.1: 34794553-34794553 on Build GRCh38
rs761437490 Chr.1: 34794532-34794532 on Build GRCh38
rs747338281 Chr.1: 34794657-34794657 on Build GRCh38
rs186793635 Chr.1: 34794620-34794620 on Build GRCh38
rs1051167 Chr.1: 34794608-34794608 on Build GRCh38
rs181609442 Chr.1: 34794597-34794597 on Build GRCh38
rs772619500 Chr.1: 34794522-34794522 on Build GRCh38
rs751546404 Chr.1: 34794564-34794564 on Build GRCh38
rs777214569 Chr.1: 34794682-34794682 on Build GRCh38
rs868582477 Chr.1: 34794659-34794659 on Build GRCh38
rs759914435 Chr.1: 34794517-34794517 on Build GRCh38
rs375786829 Chr.1: 34794536-34794538 on Build GRCh38
rs61737154 Chr.1: 34794627-34794627 on Build GRCh38
rs761133887 Chr.1: 34794526-34794526 on Build GRCh38
rs778820841 Chr.1: 34794650-34794650 on Build GRCh38
rs765204798 Chr.1: 34794531-34794531 on Build GRCh38
rs769143037 Chr.1: 34794600-34794600 on Build GRCh38
rs766289151 Chr.1: 34794614-34794614 on Build GRCh38
rs61777220 Chr.1: 34794607-34794607 on Build GRCh38
rs995796281 Chr.1: 34794563-34794563 on Build GRCh38
rs41266431 Chr.1: 34794601-34794601 on Build GRCh38
rs374035543 Chr.1: 34794652-34794652 on Build GRCh38
rs754904764 Chr.1: 34794585-34794585 on Build GRCh38
rs749486425 Chr.1: 34794486-34794486 on Build GRCh38
rs749383822 Chr.1: 34794587-34794587 on Build GRCh38
rs564420000 Chr.1: 34794654-34794654 on Build GRCh38
rs761225841 Chr.1: 34794599-34794599 on Build GRCh38
rs148827352 Chr.1: 34794637-34794637 on Build GRCh38
rs950386492 Chr.1: 34794523-34794523 on Build GRCh38
rs147128480 Chr.1: 34794596-34794596 on Build GRCh38
rs376427272 Chr.1: 34794604-34794604 on Build GRCh38
rs867814052 Chr.1: 34794554-34794554 on Build GRCh38
rs200553273 Chr.1: 34794591-34794591 on Build GRCh38
rs552638096 Chr.1: 34794590-34794590 on Build GRCh38
rs769090616 Chr.1: 34794667-34794667 on Build GRCh38
rs776003230 Chr.1: 34794525-34794525 on Build GRCh38
rs61737155 Chr.1: 34794632-34794632 on Build GRCh38
rs758561735 Chr.1: 34794656-34794656 on Build GRCh38
rs753916023 Chr.1: 34794636-34794636 on Build GRCh38

View Full Product Details