Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3148706-3148933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTKD5
Entrez Gene ID
60493
Gene Name
FAST kinase domains 5
Gene Aliases
dJ1187M17.5
UniGene
Hs.741168
Gene Chromosome Location
Chr.20: 3146519-3159886 on Build GRCh38
Transcripts
NM_021826.4 NM_001267584.1 NM_014948.3 NM_199415.2 NR_038395.1

Gene Symbol
UBOX5
Entrez Gene ID
22888
Gene Name
U-box domain containing 5
Gene Aliases
RNF37, UBCE7IP5, UIP5, hUIP5
UniGene
Hs.654646
Gene Chromosome Location
Chr.20: 3107573-3159910 on Build GRCh38
Transcripts
NM_021826.4 NM_001267584.1 NM_014948.3 NM_199415.2 NR_038395.1

Gene Symbol
UBOX5-AS1
Entrez Gene ID
100134015
Gene Name
UBOX5 antisense RNA 1
Gene Aliases
-
UniGene
Hs.129448
Gene Chromosome Location
Chr.20: 3106911-3150867 on Build GRCh38
Transcripts
NM_021826.4 NM_001267584.1 NM_014948.3 NM_199415.2 NR_038395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160988 c.337G>A p.E113K Chr.20: 3148734-3148734 on Build GRCh38
1025772 c.220G>T p.A74S Chr.20: 3148851-3148851 on Build GRCh38
7177770 c.240C>T p.F80F Chr.20: 3148831-3148831 on Build GRCh38
4399168 c.264C>T p.A88A Chr.20: 3148807-3148807 on Build GRCh38
1025769 c.347G>A p.R116Q Chr.20: 3148724-3148724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756980695 Chr.20: 3148872-3148872 on Build GRCh38
rs374950346 Chr.20: 3148847-3148847 on Build GRCh38
rs201196114 Chr.20: 3148874-3148874 on Build GRCh38
rs763651209 Chr.20: 3148822-3148822 on Build GRCh38
rs79033323 Chr.20: 3148882-3148882 on Build GRCh38
rs199708716 Chr.20: 3148785-3148785 on Build GRCh38
rs766886179 Chr.20: 3148822-3148823 on Build GRCh38
rs76392282 Chr.20: 3148795-3148795 on Build GRCh38
rs768320896 Chr.20: 3148801-3148801 on Build GRCh38
rs773953334 Chr.20: 3148844-3148844 on Build GRCh38
rs754400346 Chr.20: 3148858-3148858 on Build GRCh38
rs199920576 Chr.20: 3148867-3148867 on Build GRCh38
rs762452807 Chr.20: 3148815-3148815 on Build GRCh38
rs557602257 Chr.20: 3148851-3148851 on Build GRCh38
rs201339262 Chr.20: 3148848-3148848 on Build GRCh38
rs143277441 Chr.20: 3148782-3148782 on Build GRCh38
rs760983703 Chr.20: 3148817-3148818 on Build GRCh38
rs779099840 Chr.20: 3148786-3148786 on Build GRCh38
rs540055875 Chr.20: 3148798-3148798 on Build GRCh38
rs919661639 Chr.20: 3148894-3148894 on Build GRCh38
rs748263578 Chr.20: 3148789-3148789 on Build GRCh38
rs1041370162 Chr.20: 3148790-3148790 on Build GRCh38
rs369980933 Chr.20: 3148810-3148810 on Build GRCh38
rs200117497 Chr.20: 3148783-3148783 on Build GRCh38
rs138163389 Chr.20: 3148905-3148905 on Build GRCh38
rs750278791 Chr.20: 3148875-3148875 on Build GRCh38
rs141315308 Chr.20: 3148770-3148770 on Build GRCh38
rs774921489 Chr.20: 3148812-3148812 on Build GRCh38
rs752234966 Chr.20: 3148738-3148738 on Build GRCh38
rs143020608 Chr.20: 3148911-3148911 on Build GRCh38
rs867358582 Chr.20: 3148787-3148787 on Build GRCh38
rs145622355 Chr.20: 3148756-3148756 on Build GRCh38
rs757143824 Chr.20: 3148776-3148776 on Build GRCh38
rs780156595 Chr.20: 3148893-3148893 on Build GRCh38
rs138335304 Chr.20: 3148764-3148764 on Build GRCh38
rs900011789 Chr.20: 3148854-3148854 on Build GRCh38
rs115238082 Chr.20: 3148799-3148799 on Build GRCh38
rs764762315 Chr.20: 3148730-3148730 on Build GRCh38
rs150800870 Chr.20: 3148896-3148896 on Build GRCh38

View Full Product Details