Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 195771549-195771806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH7
Entrez Gene ID
56171
Gene Name
dynein axonemal heavy chain 7
Gene Aliases
-
UniGene
Hs.97403
Gene Chromosome Location
Chr.2: 195737604-196075188 on Build GRCh38
Transcripts
NM_018897.2 XM_011511487.2 XM_011511488.2 XM_011511489.2 XM_011511490.2 XM_011511497.2 XM_017004504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5861402 c.11383T>G p.L3795V Chr.2: 195771710-195771710 on Build GRCh38
6089699 c.11410G>T p.V3804L Chr.2: 195771683-195771683 on Build GRCh38
6575258 c.11419C>G p.Q3807E Chr.2: 195771674-195771674 on Build GRCh38
5429213 c.11332A>G p.T3778A Chr.2: 195771761-195771761 on Build GRCh38
5903391 c.11433+8G>A p.? Chr.2: 195771652-195771652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377069668 Chr.2: 195771773-195771773 on Build GRCh38
rs768232823 Chr.2: 195771692-195771692 on Build GRCh38
rs755306696 Chr.2: 195771668-195771668 on Build GRCh38
rs373836682 Chr.2: 195771778-195771778 on Build GRCh38
rs748812309 Chr.2: 195771690-195771690 on Build GRCh38
rs754107580 Chr.2: 195771652-195771652 on Build GRCh38
rs552086773 Chr.2: 195771599-195771599 on Build GRCh38
rs771950965 Chr.2: 195771715-195771715 on Build GRCh38
rs774084942 Chr.2: 195771699-195771699 on Build GRCh38
rs931490363 Chr.2: 195771666-195771666 on Build GRCh38
rs752982405 Chr.2: 195771737-195771737 on Build GRCh38
rs371038840 Chr.2: 195771775-195771775 on Build GRCh38
rs527896031 Chr.2: 195771635-195771635 on Build GRCh38
rs372501545 Chr.2: 195771750-195771750 on Build GRCh38
rs768429054 Chr.2: 195771620-195771620 on Build GRCh38
rs757069562 Chr.2: 195771634-195771635 on Build GRCh38
rs760760844 Chr.2: 195771721-195771721 on Build GRCh38
rs76167292 Chr.2: 195771634-195771634 on Build GRCh38
rs778407940 Chr.2: 195771770-195771770 on Build GRCh38
rs778864861 Chr.2: 195771635-195771635 on Build GRCh38
rs760814385 Chr.2: 195771647-195771647 on Build GRCh38
rs537879809 Chr.2: 195771598-195771598 on Build GRCh38
rs79102199 Chr.2: 195771617-195771617 on Build GRCh38
rs771903410 Chr.2: 195771776-195771777 on Build GRCh38
rs765332637 Chr.2: 195771730-195771730 on Build GRCh38
rs536343588 Chr.2: 195771684-195771684 on Build GRCh38
rs996454275 Chr.2: 195771676-195771676 on Build GRCh38
rs766551779 Chr.2: 195771724-195771724 on Build GRCh38
rs766604966 Chr.2: 195771651-195771651 on Build GRCh38
rs1001094787 Chr.2: 195771576-195771576 on Build GRCh38
rs749010468 Chr.2: 195771619-195771619 on Build GRCh38
rs765566988 Chr.2: 195771677-195771677 on Build GRCh38
rs773224567 Chr.2: 195771640-195771640 on Build GRCh38
rs867446312 Chr.2: 195771674-195771674 on Build GRCh38
rs200662508 Chr.2: 195771752-195771752 on Build GRCh38
rs765905199 Chr.2: 195771687-195771687 on Build GRCh38
rs369371792 Chr.2: 195771720-195771720 on Build GRCh38
rs59838531 Chr.2: 195771771-195771771 on Build GRCh38
rs57513754 Chr.2: 195771676-195771676 on Build GRCh38
rs764589924 Chr.2: 195771745-195771745 on Build GRCh38
rs892820321 Chr.2: 195771586-195771586 on Build GRCh38
rs747595567 Chr.2: 195771686-195771686 on Build GRCh38
rs763200107 Chr.2: 195771738-195771738 on Build GRCh38
rs753306210 Chr.2: 195771755-195771755 on Build GRCh38
rs754506919 Chr.2: 195771762-195771762 on Build GRCh38
rs369683603 Chr.2: 195771630-195771630 on Build GRCh38
rs757706559 Chr.2: 195771753-195771753 on Build GRCh38
rs534343605 Chr.2: 195771633-195771633 on Build GRCh38
rs746812162 Chr.2: 195771785-195771785 on Build GRCh38
rs769347243 Chr.2: 195771614-195771614 on Build GRCh38
rs113133988 Chr.2: 195771683-195771683 on Build GRCh38
rs759689589 Chr.2: 195771728-195771728 on Build GRCh38
rs778124013 Chr.2: 195771685-195771685 on Build GRCh38
rs745736298 Chr.2: 195771679-195771680 on Build GRCh38
rs747849544 Chr.2: 195771706-195771706 on Build GRCh38
rs556015158 Chr.2: 195771688-195771688 on Build GRCh38
rs776790815 Chr.2: 195771727-195771727 on Build GRCh38
rs530706724 Chr.2: 195771711-195771711 on Build GRCh38
rs182270960 Chr.2: 195771580-195771580 on Build GRCh38

View Full Product Details