Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75443071-75443342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD5
Entrez Gene ID
81544
Gene Name
glycerophosphodiester phosphodiesterase domain containing 5
Gene Aliases
GDE2, PP1665
UniGene
Hs.503297
Gene Chromosome Location
Chr.11: 75434640-75525916 on Build GRCh38
Transcripts
NM_030792.6 XM_006718697.3 XM_011545276.2 XM_011545277.2 XM_011545278.2 XM_011545279.2 XM_011545280.1 XM_011545281.2 XM_011545285.2 XM_011545286.1 XM_011545287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4512727 c.912C>T p.T304T Chr.11: 75443172-75443172 on Build GRCh38
4650264 c.852C>T p.N284N Chr.11: 75443232-75443232 on Build GRCh38
4574804 c.870G>A p.P290P Chr.11: 75443214-75443214 on Build GRCh38
3453355 c.816C>T p.F272F Chr.11: 75443268-75443268 on Build GRCh38
5750012 c.839G>A p.R280Q Chr.11: 75443245-75443245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368138525 Chr.11: 75443268-75443268 on Build GRCh38
rs565726247 Chr.11: 75443246-75443246 on Build GRCh38
rs938447900 Chr.11: 75443301-75443301 on Build GRCh38
rs779162166 Chr.11: 75443163-75443163 on Build GRCh38
rs755223071 Chr.11: 75443162-75443162 on Build GRCh38
rs1053002254 Chr.11: 75443144-75443144 on Build GRCh38
rs747881918 Chr.11: 75443177-75443177 on Build GRCh38
rs145858369 Chr.11: 75443159-75443159 on Build GRCh38
rs768225196 Chr.11: 75443222-75443222 on Build GRCh38
rs1006758161 Chr.11: 75443220-75443222 on Build GRCh38
rs952572099 Chr.11: 75443215-75443215 on Build GRCh38
rs746967658 Chr.11: 75443221-75443221 on Build GRCh38
rs999984207 Chr.11: 75443137-75443137 on Build GRCh38
rs904390801 Chr.11: 75443122-75443122 on Build GRCh38
rs914843488 Chr.11: 75443253-75443253 on Build GRCh38
rs371106113 Chr.11: 75443292-75443292 on Build GRCh38
rs148907682 Chr.11: 75443114-75443114 on Build GRCh38
rs143749567 Chr.11: 75443220-75443220 on Build GRCh38
rs1017026830 Chr.11: 75443200-75443200 on Build GRCh38
rs935711512 Chr.11: 75443293-75443293 on Build GRCh38
rs755062257 Chr.11: 75443242-75443242 on Build GRCh38
rs149014681 Chr.11: 75443189-75443189 on Build GRCh38
rs745329093 Chr.11: 75443280-75443280 on Build GRCh38
rs777523138 Chr.11: 75443271-75443271 on Build GRCh38
rs199987196 Chr.11: 75443206-75443206 on Build GRCh38
rs371432695 Chr.11: 75443158-75443158 on Build GRCh38
rs775928676 Chr.11: 75443265-75443271 on Build GRCh38
rs992394812 Chr.11: 75443225-75443225 on Build GRCh38
rs758141935 Chr.11: 75443249-75443249 on Build GRCh38
rs766862233 Chr.11: 75443234-75443234 on Build GRCh38
rs757217461 Chr.11: 75443276-75443276 on Build GRCh38
rs143224220 Chr.11: 75443308-75443308 on Build GRCh38
rs916932614 Chr.11: 75443231-75443231 on Build GRCh38
rs773621642 Chr.11: 75443302-75443302 on Build GRCh38
rs746824093 Chr.11: 75443273-75443273 on Build GRCh38
rs529319755 Chr.11: 75443156-75443156 on Build GRCh38
rs759037947 Chr.11: 75443204-75443204 on Build GRCh38
rs770826317 Chr.11: 75443203-75443203 on Build GRCh38
rs954056036 Chr.11: 75443187-75443187 on Build GRCh38
rs896819273 Chr.11: 75443320-75443320 on Build GRCh38
rs769457413 Chr.11: 75443290-75443290 on Build GRCh38
rs765378884 Chr.11: 75443243-75443243 on Build GRCh38
rs1051003733 Chr.11: 75443309-75443309 on Build GRCh38
rs1052900941 Chr.11: 75443296-75443296 on Build GRCh38
rs748923674 Chr.11: 75443300-75443300 on Build GRCh38
rs761355473 Chr.11: 75443318-75443319 on Build GRCh38
rs925698491 Chr.11: 75443267-75443267 on Build GRCh38
rs775283241 Chr.11: 75443214-75443214 on Build GRCh38
rs956450520 Chr.11: 75443245-75443245 on Build GRCh38
rs756094082 Chr.11: 75443157-75443157 on Build GRCh38
rs768067823 Chr.11: 75443102-75443102 on Build GRCh38
rs371832416 Chr.11: 75443277-75443277 on Build GRCh38
rs777011101 Chr.11: 75443319-75443319 on Build GRCh38
rs746989136 Chr.11: 75443202-75443202 on Build GRCh38
rs374548603 Chr.11: 75443233-75443233 on Build GRCh38
rs761002027 Chr.11: 75443304-75443304 on Build GRCh38

View Full Product Details