Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 91957116-91957385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
UniGene
Hs.500526
Gene Chromosome Location
Chr.10: 91923769-92030998 on Build GRCh38
Transcripts
NM_003972.2 XM_011540326.1 XM_011540327.2 XM_011540328.2 XM_011540329.2 XM_017016877.1 XM_017016878.1 XM_017016879.1 XM_017016880.1 XM_017016881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6067269 c.860G>T p.C287F Chr.10: 91957253-91957253 on Build GRCh38
1702761 c.842G>A p.W281* Chr.10: 91957235-91957235 on Build GRCh38
1702762 c.893C>T p.S298F Chr.10: 91957286-91957286 on Build GRCh38
921420 c.891C>A p.P297P Chr.10: 91957284-91957284 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs536198889 Chr.10: 91957325-91957325 on Build GRCh38
rs768713769 Chr.10: 91957321-91957321 on Build GRCh38
rs765051662 Chr.10: 91957328-91957328 on Build GRCh38
rs746390763 Chr.10: 91957283-91957283 on Build GRCh38
rs546593522 Chr.10: 91957174-91957174 on Build GRCh38
rs781058045 Chr.10: 91957221-91957221 on Build GRCh38
rs749006433 Chr.10: 91957318-91957319 on Build GRCh38
rs202035118 Chr.10: 91957200-91957200 on Build GRCh38
rs747772641 Chr.10: 91957230-91957230 on Build GRCh38
rs145741532 Chr.10: 91957232-91957232 on Build GRCh38
rs760863543 Chr.10: 91957311-91957311 on Build GRCh38
rs377094058 Chr.10: 91957177-91957177 on Build GRCh38
rs372186231 Chr.10: 91957354-91957354 on Build GRCh38
rs751447889 Chr.10: 91957343-91957343 on Build GRCh38
rs977617906 Chr.10: 91957348-91957348 on Build GRCh38
rs761998409 Chr.10: 91957327-91957327 on Build GRCh38
rs373136950 Chr.10: 91957339-91957339 on Build GRCh38
rs769341113 Chr.10: 91957303-91957304 on Build GRCh38
rs766397994 Chr.10: 91957340-91957340 on Build GRCh38
rs7894570 Chr.10: 91957142-91957142 on Build GRCh38
rs528929284 Chr.10: 91957210-91957210 on Build GRCh38
rs761661320 Chr.10: 91957182-91957182 on Build GRCh38
rs1042125731 Chr.10: 91957244-91957244 on Build GRCh38
rs764982528 Chr.10: 91957194-91957194 on Build GRCh38
rs867116135 Chr.10: 91957183-91957183 on Build GRCh38
rs779666789 Chr.10: 91957314-91957328 on Build GRCh38
rs750168897 Chr.10: 91957334-91957334 on Build GRCh38
rs902255570 Chr.10: 91957287-91957287 on Build GRCh38
rs1002423089 Chr.10: 91957163-91957163 on Build GRCh38
rs751026888 Chr.10: 91957212-91957212 on Build GRCh38
rs548955158 Chr.10: 91957217-91957217 on Build GRCh38
rs779369727 Chr.10: 91957267-91957267 on Build GRCh38
rs775444308 Chr.10: 91957310-91957310 on Build GRCh38
rs772563544 Chr.10: 91957301-91957301 on Build GRCh38
rs369627991 Chr.10: 91957202-91957202 on Build GRCh38

View Full Product Details