Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86477197-86477470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN1
Entrez Gene ID
51115
Gene Name
regulator of microtubule dynamics 1
Gene Aliases
CGI-90, FAM82B, RMD-1, RMD1
UniGene
Hs.145386
Gene Chromosome Location
Chr.8: 86467116-86514348 on Build GRCh38
Transcripts
NM_001286707.1 NM_001286719.1 NM_001317807.1 NM_016033.2 XM_005250944.3 XM_005250947.3 XM_005250948.3 XM_006716573.3 XM_011517090.2 XM_011517091.2 XM_011517092.2 XM_011517093.2 XM_011517094.2 XM_011517095.2 XM_011517096.2 XM_011517097.2 XM_011517098.2 XM_017013518.1 XM_017013519.1 XM_017013520.1 XM_017013521.1 XM_017013522.1 XM_017013523.1 XM_017013524.1 XM_017013525.1 XM_017013526.1 XM_017013527.1 XM_017013528.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7190440 c.730-7_751del29 p.? Chr.8: 86477303-86477331 on Build GRCh38
7190493 c.733_757del25 p.L245fs*14 Chr.8: 86477297-86477321 on Build GRCh38
1489461 c.747C>T p.H249H Chr.8: 86477307-86477307 on Build GRCh38
6431156 c.751G>T p.A251S Chr.8: 86477303-86477303 on Build GRCh38
6568894 c.730-3C>T p.? Chr.8: 86477327-86477327 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770090488 Chr.8: 86477317-86477317 on Build GRCh38
rs772294406 Chr.8: 86477355-86477356 on Build GRCh38
rs774641012 Chr.8: 86477361-86477361 on Build GRCh38
rs764043155 Chr.8: 86477269-86477269 on Build GRCh38
rs950306082 Chr.8: 86477438-86477438 on Build GRCh38
rs370162731 Chr.8: 86477261-86477261 on Build GRCh38
rs1048602667 Chr.8: 86477241-86477241 on Build GRCh38
rs750355856 Chr.8: 86477286-86477290 on Build GRCh38
rs200289866 Chr.8: 86477335-86477335 on Build GRCh38
rs34697397 Chr.8: 86477337-86477338 on Build GRCh38
rs1039971873 Chr.8: 86477297-86477297 on Build GRCh38
rs771123677 Chr.8: 86477345-86477345 on Build GRCh38
rs749862095 Chr.8: 86477327-86477327 on Build GRCh38
rs973472230 Chr.8: 86477310-86477310 on Build GRCh38
rs146063970 Chr.8: 86477304-86477304 on Build GRCh38
rs199708809 Chr.8: 86477333-86477335 on Build GRCh38
rs917403501 Chr.8: 86477402-86477402 on Build GRCh38
rs562305019 Chr.8: 86477397-86477397 on Build GRCh38
rs543394266 Chr.8: 86477232-86477232 on Build GRCh38
rs66648775 Chr.8: 86477334-86477335 on Build GRCh38
rs542507927 Chr.8: 86477381-86477381 on Build GRCh38
rs767284715 Chr.8: 86477426-86477426 on Build GRCh38
rs998359379 Chr.8: 86477416-86477416 on Build GRCh38
rs772280046 Chr.8: 86477374-86477374 on Build GRCh38
rs756045752 Chr.8: 86477301-86477302 on Build GRCh38
rs778442051 Chr.8: 86477279-86477279 on Build GRCh38
rs202043282 Chr.8: 86477337-86477337 on Build GRCh38
rs958127421 Chr.8: 86477230-86477230 on Build GRCh38
rs753804521 Chr.8: 86477270-86477270 on Build GRCh38
rs759695407 Chr.8: 86477371-86477371 on Build GRCh38
rs7461278 Chr.8: 86477322-86477322 on Build GRCh38
rs532044107 Chr.8: 86477302-86477302 on Build GRCh38
rs138797821 Chr.8: 86477320-86477320 on Build GRCh38
rs551847149 Chr.8: 86477338-86477339 on Build GRCh38
rs1009361181 Chr.8: 86477242-86477242 on Build GRCh38
rs964669145 Chr.8: 86477300-86477300 on Build GRCh38
rs750060915 Chr.8: 86477280-86477280 on Build GRCh38
rs563231943 Chr.8: 86477277-86477277 on Build GRCh38

View Full Product Details