Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54465264-54465513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42EP5
Entrez Gene ID
148170
Gene Name
CDC42 effector protein 5
Gene Aliases
Borg3, CEP5
UniGene
Hs.415791
Gene Chromosome Location
Chr.19: 54465025-54473290 on Build GRCh38
Transcripts
NM_145057.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7332203 c.206G>T p.R69L Chr.19: 54465342-54465342 on Build GRCh38
4670730 c.112C>T p.R38C Chr.19: 54465436-54465436 on Build GRCh38
4670729 c.173C>T p.P58L Chr.19: 54465375-54465375 on Build GRCh38
7300165 c.113G>A p.R38H Chr.19: 54465435-54465435 on Build GRCh38
6840930 c.118G>A p.G40S Chr.19: 54465430-54465430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780052687 Chr.19: 54465332-54465332 on Build GRCh38
rs866655876 Chr.19: 54465300-54465300 on Build GRCh38
rs749379600 Chr.19: 54465335-54465335 on Build GRCh38
rs749372033 Chr.19: 54465449-54465449 on Build GRCh38
rs773647408 Chr.19: 54465408-54465408 on Build GRCh38
rs1032633481 Chr.19: 54465306-54465306 on Build GRCh38
rs768796493 Chr.19: 54465339-54465339 on Build GRCh38
rs759919377 Chr.19: 54465490-54465490 on Build GRCh38
rs773792222 Chr.19: 54465496-54465496 on Build GRCh38
rs901032821 Chr.19: 54465340-54465340 on Build GRCh38
rs780924608 Chr.19: 54465314-54465314 on Build GRCh38
rs766898419 Chr.19: 54465419-54465419 on Build GRCh38
rs774607002 Chr.19: 54465364-54465364 on Build GRCh38
rs780977544 Chr.19: 54465440-54465440 on Build GRCh38
rs201220343 Chr.19: 54465461-54465461 on Build GRCh38
rs778822762 Chr.19: 54465460-54465460 on Build GRCh38
rs930680419 Chr.19: 54465383-54465383 on Build GRCh38
rs745808060 Chr.19: 54465318-54465318 on Build GRCh38
rs750204751 Chr.19: 54465443-54465443 on Build GRCh38
rs752195595 Chr.19: 54465289-54465289 on Build GRCh38
rs773308660 Chr.19: 54465446-54465447 on Build GRCh38
rs761022040 Chr.19: 54465495-54465496 on Build GRCh38
rs771384155 Chr.19: 54465482-54465482 on Build GRCh38
rs751242744 Chr.19: 54465436-54465436 on Build GRCh38
rs773771994 Chr.19: 54465467-54465467 on Build GRCh38
rs969742102 Chr.19: 54465288-54465288 on Build GRCh38
rs752217646 Chr.19: 54465430-54465430 on Build GRCh38
rs974915124 Chr.19: 54465374-54465374 on Build GRCh38
rs895702369 Chr.19: 54465477-54465477 on Build GRCh38
rs758046869 Chr.19: 54465294-54465294 on Build GRCh38
rs944966996 Chr.19: 54465350-54465350 on Build GRCh38
rs768806333 Chr.19: 54465450-54465450 on Build GRCh38
rs772600002 Chr.19: 54465398-54465398 on Build GRCh38
rs756108701 Chr.19: 54465445-54465445 on Build GRCh38
rs756836127 Chr.19: 54465437-54465437 on Build GRCh38
rs780007502 Chr.19: 54465446-54465446 on Build GRCh38
rs761280702 Chr.19: 54465411-54465411 on Build GRCh38
rs1005515652 Chr.19: 54465382-54465382 on Build GRCh38
rs762493539 Chr.19: 54465434-54465434 on Build GRCh38
rs541272939 Chr.19: 54465313-54465313 on Build GRCh38
rs1028382389 Chr.19: 54465366-54465366 on Build GRCh38
rs747374305 Chr.19: 54465476-54465476 on Build GRCh38
rs772366145 Chr.19: 54465466-54465466 on Build GRCh38
rs769659275 Chr.19: 54465329-54465329 on Build GRCh38
rs17856722 Chr.19: 54465373-54465373 on Build GRCh38
rs763663770 Chr.19: 54465493-54465493 on Build GRCh38
rs763704316 Chr.19: 54465435-54465435 on Build GRCh38
rs751351879 Chr.19: 54465295-54465295 on Build GRCh38
rs777175682 Chr.19: 54465488-54465488 on Build GRCh38
rs998205581 Chr.19: 54465355-54465355 on Build GRCh38
rs557776710 Chr.19: 54465395-54465395 on Build GRCh38
rs922096963 Chr.19: 54465379-54465379 on Build GRCh38
rs866337782 Chr.19: 54465292-54465292 on Build GRCh38
rs912177413 Chr.19: 54465349-54465349 on Build GRCh38

View Full Product Details