Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150467153-150467384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP8
Entrez Gene ID
155038
Gene Name
GTPase, IMAP family member 8
Gene Aliases
IAN-9, IAN6, IAN9, IANT
UniGene
Hs.647121
Gene Chromosome Location
Chr.7: 150450630-150479395 on Build GRCh38
Transcripts
NM_175571.3 XM_005249950.4 XM_005249951.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4924874 c.481T>A p.Y161N Chr.7: 150467179-150467179 on Build GRCh38
3635936 c.555C>T p.L185L Chr.7: 150467253-150467253 on Build GRCh38
1699556 c.526G>A p.D176N Chr.7: 150467224-150467224 on Build GRCh38
7369457 c.634C>A p.Q212K Chr.7: 150467332-150467332 on Build GRCh38
3635935 c.537C>T p.I179I Chr.7: 150467235-150467235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540607407 Chr.7: 150467257-150467257 on Build GRCh38
rs759897976 Chr.7: 150467207-150467207 on Build GRCh38
rs751000509 Chr.7: 150467229-150467229 on Build GRCh38
rs758616514 Chr.7: 150467262-150467262 on Build GRCh38
rs202178130 Chr.7: 150467218-150467218 on Build GRCh38
rs749750785 Chr.7: 150467263-150467263 on Build GRCh38
rs771211629 Chr.7: 150467191-150467191 on Build GRCh38
rs200592029 Chr.7: 150467258-150467258 on Build GRCh38
rs763584946 Chr.7: 150467227-150467227 on Build GRCh38
rs755917939 Chr.7: 150467317-150467317 on Build GRCh38
rs750066764 Chr.7: 150467300-150467300 on Build GRCh38
rs761270896 Chr.7: 150467295-150467295 on Build GRCh38
rs756911801 Chr.7: 150467232-150467232 on Build GRCh38
rs201302071 Chr.7: 150467220-150467220 on Build GRCh38
rs776560216 Chr.7: 150467245-150467245 on Build GRCh38
rs866765845 Chr.7: 150467224-150467224 on Build GRCh38
rs145618535 Chr.7: 150467180-150467180 on Build GRCh38
rs771081911 Chr.7: 150467276-150467276 on Build GRCh38
rs753219773 Chr.7: 150467215-150467215 on Build GRCh38
rs147231393 Chr.7: 150467287-150467287 on Build GRCh38
rs780846275 Chr.7: 150467241-150467241 on Build GRCh38
rs756123094 Chr.7: 150467189-150467189 on Build GRCh38
rs762501861 Chr.7: 150467217-150467217 on Build GRCh38
rs764523607 Chr.7: 150467290-150467290 on Build GRCh38
rs778005273 Chr.7: 150467273-150467273 on Build GRCh38
rs765695742 Chr.7: 150467208-150467208 on Build GRCh38
rs116316852 Chr.7: 150467254-150467254 on Build GRCh38
rs760973666 Chr.7: 150467188-150467188 on Build GRCh38
rs753620580 Chr.7: 150467344-150467344 on Build GRCh38
rs767113572 Chr.7: 150467299-150467299 on Build GRCh38
rs776873502 Chr.7: 150467282-150467282 on Build GRCh38
rs753025993 Chr.7: 150467279-150467279 on Build GRCh38
rs770174770 Chr.7: 150467286-150467286 on Build GRCh38
rs746261806 Chr.7: 150467284-150467284 on Build GRCh38
rs747028717 Chr.7: 150467275-150467275 on Build GRCh38
rs766191571 Chr.7: 150467332-150467332 on Build GRCh38
rs776872577 Chr.7: 150467205-150467205 on Build GRCh38
rs763450247 Chr.7: 150467289-150467289 on Build GRCh38
rs758169110 Chr.7: 150467202-150467204 on Build GRCh38
rs773869620 Chr.7: 150467293-150467293 on Build GRCh38
rs754981356 Chr.7: 150467347-150467347 on Build GRCh38
rs138233370 Chr.7: 150467350-150467350 on Build GRCh38

View Full Product Details