Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 11580671-11580944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GREB1
Entrez Gene ID
9687
Gene Name
growth regulation by estrogen in breast cancer 1
Gene Aliases
-
UniGene
Hs.467733
Gene Chromosome Location
Chr.2: 11534116-11642788 on Build GRCh38
Transcripts
NM_014668.3 NM_033090.2 NM_148903.2 XM_005246192.4 XM_011510418.2 XM_011510419.2 XM_011510423.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6841243 c.886C>T p.R296* Chr.2: 11580817-11580817 on Build GRCh38
3040749 c.887G>T p.R296L Chr.2: 11580818-11580818 on Build GRCh38
441038 c.825C>T p.F275F Chr.2: 11580756-11580756 on Build GRCh38
275371 c.864G>T p.K288N Chr.2: 11580795-11580795 on Build GRCh38
6257642 c.877G>T p.A293S Chr.2: 11580808-11580808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764330178 Chr.2: 11580726-11580726 on Build GRCh38
rs201637674 Chr.2: 11580810-11580810 on Build GRCh38
rs750847838 Chr.2: 11580738-11580738 on Build GRCh38
rs776554870 Chr.2: 11580898-11580898 on Build GRCh38
rs1020420883 Chr.2: 11580767-11580767 on Build GRCh38
rs376693873 Chr.2: 11580740-11580740 on Build GRCh38
rs200145343 Chr.2: 11580854-11580854 on Build GRCh38
rs752071514 Chr.2: 11580825-11580825 on Build GRCh38
rs768624823 Chr.2: 11580815-11580815 on Build GRCh38
rs766482147 Chr.2: 11580693-11580693 on Build GRCh38
rs760875680 Chr.2: 11580722-11580722 on Build GRCh38
rs894007741 Chr.2: 11580770-11580770 on Build GRCh38
rs139908729 Chr.2: 11580924-11580924 on Build GRCh38
rs76718398 Chr.2: 11580760-11580760 on Build GRCh38
rs13004648 Chr.2: 11580860-11580860 on Build GRCh38
rs201070918 Chr.2: 11580824-11580824 on Build GRCh38
rs749143603 Chr.2: 11580779-11580779 on Build GRCh38
rs150918268 Chr.2: 11580759-11580759 on Build GRCh38
rs931696866 Chr.2: 11580800-11580800 on Build GRCh38
rs149713953 Chr.2: 11580712-11580712 on Build GRCh38
rs757575403 Chr.2: 11580732-11580732 on Build GRCh38
rs199738668 Chr.2: 11580773-11580773 on Build GRCh38
rs371267216 Chr.2: 11580878-11580878 on Build GRCh38
rs770864047 Chr.2: 11580874-11580874 on Build GRCh38
rs1050321012 Chr.2: 11580743-11580743 on Build GRCh38
rs770958241 Chr.2: 11580783-11580783 on Build GRCh38
rs774460720 Chr.2: 11580787-11580787 on Build GRCh38
rs981862488 Chr.2: 11580922-11580922 on Build GRCh38
rs973636713 Chr.2: 11580872-11580872 on Build GRCh38
rs780581197 Chr.2: 11580745-11580745 on Build GRCh38
rs148901217 Chr.2: 11580746-11580746 on Build GRCh38
rs749050571 Chr.2: 11580873-11580873 on Build GRCh38
rs765549350 Chr.2: 11580821-11580821 on Build GRCh38
rs778152733 Chr.2: 11580756-11580756 on Build GRCh38
rs866862670 Chr.2: 11580814-11580814 on Build GRCh38
rs778928025 Chr.2: 11580893-11580893 on Build GRCh38
rs762978702 Chr.2: 11580692-11580692 on Build GRCh38
rs777608716 Chr.2: 11580861-11580861 on Build GRCh38
rs368334075 Chr.2: 11580747-11580747 on Build GRCh38
rs772106391 Chr.2: 11580896-11580896 on Build GRCh38
rs147997301 Chr.2: 11580734-11580734 on Build GRCh38
rs551258797 Chr.2: 11580921-11580921 on Build GRCh38
rs761928277 Chr.2: 11580817-11580817 on Build GRCh38
rs183530916 Chr.2: 11580853-11580853 on Build GRCh38
rs899920855 Chr.2: 11580785-11580785 on Build GRCh38
rs779571742 Chr.2: 11580816-11580816 on Build GRCh38
rs750759720 Chr.2: 11580822-11580822 on Build GRCh38
rs1039831045 Chr.2: 11580780-11580780 on Build GRCh38
rs760781224 Chr.2: 11580792-11580792 on Build GRCh38
rs16857665 Chr.2: 11580774-11580774 on Build GRCh38
rs763342408 Chr.2: 11580823-11580823 on Build GRCh38

View Full Product Details