Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 150567641-150567910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2A
Entrez Gene ID
83939
Gene Name
eukaryotic translation initiation factor 2A
Gene Aliases
CDA02, EIF-2A, MST089, MSTP004, MSTP089
UniGene
Hs.655782
Gene Chromosome Location
Chr.3: 150546678-150586016 on Build GRCh38
Transcripts
NM_001319043.1 NM_001319044.1 NM_001319045.1 NM_001319046.1 NM_032025.4 XM_011513224.1 XM_011512698.2

Gene Symbol
SERP1
Entrez Gene ID
27230
Gene Name
stress-associated endoplasmic reticulum protein 1
Gene Aliases
RAMP4
UniGene
Hs.518326
Gene Chromosome Location
Chr.3: 150541993-150603177 on Build GRCh38
Transcripts
NM_001319043.1 NM_001319044.1 NM_001319045.1 NM_001319046.1 NM_032025.4 XM_011513224.1 XM_011512698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1419891 c.431A>C p.K144T Chr.3: 150567723-150567723 on Build GRCh38
3729826 c.401-10delT p.? Chr.3: 150567683-150567683 on Build GRCh38
7417098 c.436A>T p.N146Y Chr.3: 150567728-150567728 on Build GRCh38
5701252 c.503_504insA p.I170fs*2 Chr.3: 150567720-150567721 on Build GRCh38
5701253 c.428_429insA p.I145fs*2 Chr.3: 150567720-150567721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190253236 Chr.3: 150567790-150567790 on Build GRCh38
rs774407654 Chr.3: 150567737-150567737 on Build GRCh38
rs766386697 Chr.3: 150567791-150567791 on Build GRCh38
rs373435793 Chr.3: 150567736-150567736 on Build GRCh38
rs756441893 Chr.3: 150567697-150567697 on Build GRCh38
rs753181208 Chr.3: 150567864-150567864 on Build GRCh38
rs548363236 Chr.3: 150567833-150567833 on Build GRCh38
rs768111338 Chr.3: 150567809-150567809 on Build GRCh38
rs527839208 Chr.3: 150567870-150567870 on Build GRCh38
rs564329374 Chr.3: 150567754-150567754 on Build GRCh38
rs747738773 Chr.3: 150567680-150567681 on Build GRCh38
rs755472438 Chr.3: 150567803-150567803 on Build GRCh38
rs866479856 Chr.3: 150567747-150567747 on Build GRCh38
rs778585415 Chr.3: 150567707-150567707 on Build GRCh38
rs751611547 Chr.3: 150567792-150567792 on Build GRCh38
rs929098132 Chr.3: 150567693-150567693 on Build GRCh38
rs748473134 Chr.3: 150567690-150567690 on Build GRCh38
rs765098331 Chr.3: 150567777-150567777 on Build GRCh38
rs139559220 Chr.3: 150567774-150567774 on Build GRCh38
rs769244111 Chr.3: 150567682-150567683 on Build GRCh38
rs2293150 Chr.3: 150567877-150567877 on Build GRCh38
rs376872417 Chr.3: 150567748-150567748 on Build GRCh38
rs371505750 Chr.3: 150567757-150567757 on Build GRCh38
rs917674798 Chr.3: 150567672-150567672 on Build GRCh38
rs533264835 Chr.3: 150567760-150567760 on Build GRCh38
rs2293151 Chr.3: 150567733-150567733 on Build GRCh38
rs956143709 Chr.3: 150567846-150567846 on Build GRCh38
rs562721762 Chr.3: 150567784-150567784 on Build GRCh38
rs777308633 Chr.3: 150567732-150567733 on Build GRCh38
rs766931871 Chr.3: 150567800-150567800 on Build GRCh38
rs900315395 Chr.3: 150567669-150567669 on Build GRCh38
rs878927513 Chr.3: 150567790-150567791 on Build GRCh38
rs749233731 Chr.3: 150567779-150567782 on Build GRCh38
rs373948705 Chr.3: 150567878-150567878 on Build GRCh38
rs568653366 Chr.3: 150567836-150567836 on Build GRCh38
rs960183780 Chr.3: 150567848-150567848 on Build GRCh38
rs1048032752 Chr.3: 150567694-150567694 on Build GRCh38

View Full Product Details