Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33172705-33172978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA2
Entrez Gene ID
140683
Gene Name
BPI fold containing family A member 2
Gene Aliases
C20orf70, PSP, SPLUNC2, bA49G10.1
UniGene
Hs.125960
Gene Chromosome Location
Chr.20: 33161848-33181417 on Build GRCh38
Transcripts
NM_001319164.1 NM_080574.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
140260 c.181G>A p.D61N Chr.20: 33172955-33172955 on Build GRCh38
3379018 c.190G>A p.V64M Chr.20: 33172964-33172964 on Build GRCh38
3405025 c.163C>A p.L55I Chr.20: 33172937-33172937 on Build GRCh38
5391840 c.174G>A p.L58L Chr.20: 33172948-33172948 on Build GRCh38
4401222 c.189A>G p.G63G Chr.20: 33172963-33172963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865888709 Chr.20: 33172865-33172865 on Build GRCh38
rs58172152 Chr.20: 33172954-33172954 on Build GRCh38
rs566694836 Chr.20: 33172745-33172745 on Build GRCh38
rs751528374 Chr.20: 33172947-33172947 on Build GRCh38
rs890462129 Chr.20: 33172937-33172937 on Build GRCh38
rs762415567 Chr.20: 33172927-33172927 on Build GRCh38
rs948993493 Chr.20: 33172843-33172843 on Build GRCh38
rs993963696 Chr.20: 33172775-33172775 on Build GRCh38
rs6579083 Chr.20: 33172902-33172902 on Build GRCh38
rs200080455 Chr.20: 33172929-33172929 on Build GRCh38
rs1005773455 Chr.20: 33172727-33172727 on Build GRCh38
rs774761077 Chr.20: 33172922-33172922 on Build GRCh38
rs769184439 Chr.20: 33172917-33172917 on Build GRCh38
rs1050403941 Chr.20: 33172935-33172935 on Build GRCh38
rs896862740 Chr.20: 33172755-33172755 on Build GRCh38
rs774024155 Chr.20: 33172891-33172891 on Build GRCh38
rs117186940 Chr.20: 33172744-33172744 on Build GRCh38
rs760819132 Chr.20: 33172952-33172952 on Build GRCh38
rs929560478 Chr.20: 33172761-33172761 on Build GRCh38
rs908205066 Chr.20: 33172794-33172794 on Build GRCh38
rs75887969 Chr.20: 33172940-33172940 on Build GRCh38
rs544639967 Chr.20: 33172862-33172862 on Build GRCh38
rs185238711 Chr.20: 33172776-33172776 on Build GRCh38
rs868594602 Chr.20: 33172795-33172795 on Build GRCh38
rs373945508 Chr.20: 33172932-33172932 on Build GRCh38
rs578221463 Chr.20: 33172879-33172879 on Build GRCh38
rs911098339 Chr.20: 33172949-33172949 on Build GRCh38
rs777652895 Chr.20: 33172899-33172899 on Build GRCh38
rs990692046 Chr.20: 33172886-33172886 on Build GRCh38
rs374774599 Chr.20: 33172955-33172955 on Build GRCh38
rs747949166 Chr.20: 33172890-33172890 on Build GRCh38
rs746603996 Chr.20: 33172901-33172901 on Build GRCh38
rs555908080 Chr.20: 33172819-33172819 on Build GRCh38
rs546969309 Chr.20: 33172916-33172916 on Build GRCh38
rs542770426 Chr.20: 33172944-33172944 on Build GRCh38
rs1046193324 Chr.20: 33172856-33172856 on Build GRCh38
rs971074262 Chr.20: 33172822-33172822 on Build GRCh38
rs776280895 Chr.20: 33172903-33172903 on Build GRCh38
rs574004778 Chr.20: 33172855-33172855 on Build GRCh38
rs140852113 Chr.20: 33172864-33172864 on Build GRCh38

View Full Product Details