Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87322629-87322884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B13
Entrez Gene ID
345275
Gene Name
hydroxysteroid 17-beta dehydrogenase 13
Gene Aliases
HMFN0376, NIIL497, SCDR9, SDR16C3
UniGene
Hs.284414
Gene Chromosome Location
Chr.4: 87303789-87322906 on Build GRCh38
Transcripts
NM_001136230.2 NM_178135.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4126569 c.182G>A p.S61N Chr.4: 87322660-87322660 on Build GRCh38
6168373 c.58G>C p.E20Q Chr.4: 87322784-87322784 on Build GRCh38
5355826 c.75T>G p.F25L Chr.4: 87322767-87322767 on Build GRCh38
6192938 c.63G>A p.S21S Chr.4: 87322779-87322779 on Build GRCh38
6192939 c.43A>C p.I15L Chr.4: 87322799-87322799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370077116 Chr.4: 87322861-87322861 on Build GRCh38
rs973333376 Chr.4: 87322739-87322739 on Build GRCh38
rs765811976 Chr.4: 87322683-87322683 on Build GRCh38
rs751707061 Chr.4: 87322786-87322786 on Build GRCh38
rs373745664 Chr.4: 87322705-87322705 on Build GRCh38
rs893550931 Chr.4: 87322841-87322841 on Build GRCh38
rs201312897 Chr.4: 87322732-87322732 on Build GRCh38
rs769644963 Chr.4: 87322776-87322778 on Build GRCh38
rs772966438 Chr.4: 87322753-87322753 on Build GRCh38
rs764572652 Chr.4: 87322784-87322784 on Build GRCh38
rs755020585 Chr.4: 87322837-87322837 on Build GRCh38
rs370787852 Chr.4: 87322710-87322710 on Build GRCh38
rs535092394 Chr.4: 87322745-87322745 on Build GRCh38
rs1052570727 Chr.4: 87322814-87322814 on Build GRCh38
rs781391869 Chr.4: 87322729-87322729 on Build GRCh38
rs753498673 Chr.4: 87322861-87322862 on Build GRCh38
rs137887488 Chr.4: 87322680-87322680 on Build GRCh38
rs747646094 Chr.4: 87322744-87322744 on Build GRCh38
rs763384411 Chr.4: 87322780-87322780 on Build GRCh38
rs753122915 Chr.4: 87322698-87322698 on Build GRCh38
rs1035712584 Chr.4: 87322666-87322666 on Build GRCh38
rs748220816 Chr.4: 87322834-87322834 on Build GRCh38
rs778987472 Chr.4: 87322852-87322852 on Build GRCh38
rs767778758 Chr.4: 87322795-87322795 on Build GRCh38
rs766088195 Chr.4: 87322829-87322829 on Build GRCh38
rs141927505 Chr.4: 87322675-87322675 on Build GRCh38
rs200847277 Chr.4: 87322796-87322796 on Build GRCh38
rs200528299 Chr.4: 87322664-87322664 on Build GRCh38
rs751582449 Chr.4: 87322717-87322717 on Build GRCh38
rs768568943 Chr.4: 87322736-87322736 on Build GRCh38
rs778576595 Chr.4: 87322741-87322741 on Build GRCh38
rs150242069 Chr.4: 87322820-87322820 on Build GRCh38
rs59739874 Chr.4: 87322791-87322791 on Build GRCh38
rs777457925 Chr.4: 87322794-87322795 on Build GRCh38
rs199826420 Chr.4: 87322672-87322672 on Build GRCh38
rs752295562 Chr.4: 87322727-87322730 on Build GRCh38
rs893466136 Chr.4: 87322815-87322815 on Build GRCh38
rs750261905 Chr.4: 87322730-87322730 on Build GRCh38
rs376596440 Chr.4: 87322667-87322667 on Build GRCh38
rs753608854 Chr.4: 87322835-87322835 on Build GRCh38
rs757535191 Chr.4: 87322718-87322718 on Build GRCh38
rs147733873 Chr.4: 87322779-87322779 on Build GRCh38
rs571845258 Chr.4: 87322774-87322774 on Build GRCh38
rs770076262 Chr.4: 87322775-87322775 on Build GRCh38
rs755843877 Chr.4: 87322731-87322731 on Build GRCh38

View Full Product Details