Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 111209791-111210062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF7
Entrez Gene ID
8874
Gene Name
Rho guanine nucleotide exchange factor 7
Gene Aliases
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
UniGene
Hs.508738
Gene Chromosome Location
Chr.13: 111114619-111305734 on Build GRCh38
Transcripts
NM_001113511.2 NM_001113512.2 NM_001113513.1 NM_001320851.1 NM_001320852.1 NM_001320853.1 NM_003899.3 NM_145735.3 XM_006719956.3 XM_011521133.2 XM_017020814.1 XM_017020815.1 XM_017020816.1 XM_017020817.1 XM_017020818.1 XM_017020819.1 XM_017020820.1 XM_017020821.1 XM_017020822.1 XM_017020823.1 XM_017020824.1 XM_017020825.1 XM_017020826.1 XM_017020827.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4440356 c.449G>A p.R150H Chr.13: 111209920-111209920 on Build GRCh38
6604432 c.430C>T p.R144W Chr.13: 111209964-111209964 on Build GRCh38
5006754 c.380C>T p.S127F Chr.13: 111209914-111209914 on Build GRCh38
5942521 c.437C>T p.P146L Chr.13: 111209908-111209908 on Build GRCh38
6604426 c.342C>T p.I114I Chr.13: 111209876-111209876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778628269 Chr.13: 111209963-111209963 on Build GRCh38
rs368325547 Chr.13: 111209850-111209850 on Build GRCh38
rs1031245527 Chr.13: 111209912-111209912 on Build GRCh38
rs772066171 Chr.13: 111209987-111209987 on Build GRCh38
rs1055448312 Chr.13: 111209927-111209927 on Build GRCh38
rs780802486 Chr.13: 111209860-111209860 on Build GRCh38
rs146715616 Chr.13: 111209997-111209997 on Build GRCh38
rs374730263 Chr.13: 111209991-111209991 on Build GRCh38
rs761106966 Chr.13: 111209845-111209846 on Build GRCh38
rs746348009 Chr.13: 111209856-111209856 on Build GRCh38
rs368036359 Chr.13: 111209940-111209940 on Build GRCh38
rs765660092 Chr.13: 111209909-111209909 on Build GRCh38
rs765713009 Chr.13: 111210017-111210017 on Build GRCh38
rs1050308327 Chr.13: 111209825-111209825 on Build GRCh38
rs750993031 Chr.13: 111209911-111209911 on Build GRCh38
rs776753220 Chr.13: 111210005-111210005 on Build GRCh38
rs762081813 Chr.13: 111210015-111210015 on Build GRCh38
rs762262563 Chr.13: 111209919-111209919 on Build GRCh38
rs973377476 Chr.13: 111209889-111209889 on Build GRCh38
rs754226005 Chr.13: 111209833-111209833 on Build GRCh38
rs186440416 Chr.13: 111209964-111209964 on Build GRCh38
rs267603759 Chr.13: 111209894-111209894 on Build GRCh38
rs762128521 Chr.13: 111209908-111209908 on Build GRCh38
rs775368431 Chr.13: 111209902-111209902 on Build GRCh38
rs752211697 Chr.13: 111209922-111209922 on Build GRCh38
rs777340204 Chr.13: 111209939-111209939 on Build GRCh38
rs372511961 Chr.13: 111209858-111209858 on Build GRCh38
rs760806277 Chr.13: 111209904-111209904 on Build GRCh38
rs371174541 Chr.13: 111209942-111209942 on Build GRCh38
rs1008651040 Chr.13: 111209854-111209854 on Build GRCh38
rs888862039 Chr.13: 111209838-111209838 on Build GRCh38
rs764281725 Chr.13: 111209905-111209905 on Build GRCh38
rs750681770 Chr.13: 111209973-111209973 on Build GRCh38
rs767811644 Chr.13: 111209954-111209954 on Build GRCh38
rs532466192 Chr.13: 111209821-111209821 on Build GRCh38
rs200107407 Chr.13: 111209994-111209994 on Build GRCh38
rs760717132 Chr.13: 111209995-111209995 on Build GRCh38
rs755632679 Chr.13: 111209934-111209934 on Build GRCh38
rs772154506 Chr.13: 111209900-111209900 on Build GRCh38
rs772912273 Chr.13: 111209877-111209877 on Build GRCh38
rs764765261 Chr.13: 111210023-111210024 on Build GRCh38
rs866989377 Chr.13: 111209901-111209901 on Build GRCh38
rs528095247 Chr.13: 111209820-111209820 on Build GRCh38
rs962002467 Chr.13: 111210034-111210034 on Build GRCh38
rs756235192 Chr.13: 111209826-111209826 on Build GRCh38
rs774308780 Chr.13: 111209895-111209895 on Build GRCh38
rs143399583 Chr.13: 111209950-111209950 on Build GRCh38
rs373872938 Chr.13: 111209880-111209880 on Build GRCh38
rs780851372 Chr.13: 111209920-111209920 on Build GRCh38
rs757632582 Chr.13: 111209845-111209845 on Build GRCh38
rs917781062 Chr.13: 111209941-111209941 on Build GRCh38
rs538515190 Chr.13: 111209876-111209876 on Build GRCh38
rs376311283 Chr.13: 111209832-111209832 on Build GRCh38
rs942093251 Chr.13: 111209881-111209881 on Build GRCh38
rs1023806699 Chr.13: 111209948-111209948 on Build GRCh38

View Full Product Details