Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20807318-20807543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4G3
Entrez Gene ID
8672
Gene Name
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases
eIF-4G 3, eIF4G 3, eIF4GII
UniGene
Hs.467084
Gene Chromosome Location
Chr.1: 20806292-21177320 on Build GRCh38
Transcripts
NM_001198801.1 NM_001198802.1 NM_003760.4 XM_011542356.2 XM_011542361.2 XM_011542370.1 XM_011542371.1 XM_011542374.1 XM_011542375.2 XM_011542381.2 XM_017002672.1 XM_017002673.1 XM_017002674.1 XM_017002675.1 XM_017002676.1 XM_017002677.1 XM_017002678.1 XM_017002679.1 XM_017002680.1 XM_017002681.1 XM_017002682.1 XM_017002683.1 XM_017002684.1 XM_017002685.1 XM_017002686.1 XM_017002687.1 XM_017002688.1 XM_017002689.1 XM_017002690.1 XM_017002691.1 XM_017002692.1 XM_017002693.1 XM_017002694.1 XM_017002695.1 XM_017002696.1 XM_017002697.1 XM_017002698.1 XM_017002699.1 XM_017002700.1 XM_017002701.1 XM_017002702.1 XM_017002703.1 XM_017002704.1 XM_017002705.1 XM_017002706.1 XM_017002707.1 XM_017002708.1 XM_017002709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5025201 c.4682C>A p.P1561H Chr.1: 20807413-20807413 on Build GRCh38
6323387 c.3082G>A p.E1028K Chr.1: 20807465-20807465 on Build GRCh38
7136417 c.4722C>A p.V1574V Chr.1: 20807373-20807373 on Build GRCh38
463884 c.4703G>A p.G1568D Chr.1: 20807392-20807392 on Build GRCh38
6653029 c.3189G>A p.T1063T Chr.1: 20807358-20807358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867114309 Chr.1: 20807369-20807369 on Build GRCh38
rs747687791 Chr.1: 20807505-20807507 on Build GRCh38
rs772236365 Chr.1: 20807417-20807417 on Build GRCh38
rs749034314 Chr.1: 20807359-20807359 on Build GRCh38
rs762356088 Chr.1: 20807371-20807371 on Build GRCh38
rs746060850 Chr.1: 20807383-20807383 on Build GRCh38
rs759165740 Chr.1: 20807472-20807472 on Build GRCh38
rs573551412 Chr.1: 20807430-20807430 on Build GRCh38
rs775631377 Chr.1: 20807418-20807418 on Build GRCh38
rs11584562 Chr.1: 20807403-20807403 on Build GRCh38
rs866320837 Chr.1: 20807377-20807377 on Build GRCh38
rs200368420 Chr.1: 20807406-20807406 on Build GRCh38
rs1037917085 Chr.1: 20807486-20807486 on Build GRCh38
rs765620407 Chr.1: 20807460-20807460 on Build GRCh38
rs146742995 Chr.1: 20807451-20807451 on Build GRCh38
rs770874174 Chr.1: 20807370-20807370 on Build GRCh38
rs371347072 Chr.1: 20807390-20807390 on Build GRCh38
rs540921938 Chr.1: 20807466-20807466 on Build GRCh38
rs762113871 Chr.1: 20807457-20807457 on Build GRCh38
rs767142331 Chr.1: 20807498-20807498 on Build GRCh38
rs769137304 Chr.1: 20807447-20807447 on Build GRCh38

View Full Product Details