Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 30885345-30885618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COCH
Entrez Gene ID
1690
Gene Name
cochlin
Gene Aliases
COCH-5B2, COCH5B2, DFNA9
UniGene
Hs.21016
Gene Chromosome Location
Chr.14: 30874496-30890618 on Build GRCh38
Transcripts
NM_001135058.1 NM_004086.2 XM_017021071.1 XM_017021072.1 XM_017021073.1 NR_038356.1

Gene Symbol
LOC100506071
Entrez Gene ID
100506071
Gene Name
uncharacterized LOC100506071
Gene Aliases
-
UniGene
Hs.729195
Gene Chromosome Location
Chr.14: 30876179-30889808 on Build GRCh38
Transcripts
NM_001135058.1 NM_004086.2 XM_017021071.1 XM_017021072.1 XM_017021073.1 NR_038356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3955980 c.386C>A p.P129H Chr.14: 30885493-30885493 on Build GRCh38
1201741 c.899C>A p.S300Y Chr.14: 30885559-30885559 on Build GRCh38
3495644 c.798C>T p.I266I Chr.14: 30885458-30885458 on Build GRCh38
3495646 c.929G>A p.G310E Chr.14: 30885589-30885589 on Build GRCh38
4775902 c.918_937del20 p.E307fs*6 Chr.14: 30885578-30885597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186875564 Chr.14: 30885474-30885474 on Build GRCh38
rs761471633 Chr.14: 30885471-30885471 on Build GRCh38
rs777855403 Chr.14: 30885556-30885556 on Build GRCh38
rs749516210 Chr.14: 30885400-30885400 on Build GRCh38
rs772516874 Chr.14: 30885431-30885431 on Build GRCh38
rs774715234 Chr.14: 30885409-30885409 on Build GRCh38
rs769965470 Chr.14: 30885385-30885385 on Build GRCh38
rs550268175 Chr.14: 30885552-30885552 on Build GRCh38
rs759845948 Chr.14: 30885420-30885420 on Build GRCh38
rs746994191 Chr.14: 30885376-30885376 on Build GRCh38
rs149342451 Chr.14: 30885587-30885587 on Build GRCh38
rs751540613 Chr.14: 30885523-30885523 on Build GRCh38
rs974589057 Chr.14: 30885445-30885446 on Build GRCh38
rs988785887 Chr.14: 30885379-30885379 on Build GRCh38
rs77074957 Chr.14: 30885472-30885472 on Build GRCh38
rs755026676 Chr.14: 30885538-30885538 on Build GRCh38
rs531508748 Chr.14: 30885465-30885465 on Build GRCh38
rs368609319 Chr.14: 30885506-30885506 on Build GRCh38
rs558120815 Chr.14: 30885572-30885572 on Build GRCh38
rs946045347 Chr.14: 30885553-30885553 on Build GRCh38
rs28362775 Chr.14: 30885501-30885501 on Build GRCh38
rs375612298 Chr.14: 30885446-30885446 on Build GRCh38
rs761209524 Chr.14: 30885454-30885454 on Build GRCh38
rs757456469 Chr.14: 30885559-30885559 on Build GRCh38
rs750185209 Chr.14: 30885487-30885487 on Build GRCh38
rs759202794 Chr.14: 30885388-30885388 on Build GRCh38
rs778069510 Chr.14: 30885398-30885398 on Build GRCh38
rs370194438 Chr.14: 30885404-30885404 on Build GRCh38
rs150207791 Chr.14: 30885569-30885569 on Build GRCh38
rs138805494 Chr.14: 30885578-30885578 on Build GRCh38
rs763588874 Chr.14: 30885456-30885456 on Build GRCh38
rs749336302 Chr.14: 30885558-30885558 on Build GRCh38
rs779021334 Chr.14: 30885560-30885560 on Build GRCh38
rs913285663 Chr.14: 30885510-30885510 on Build GRCh38
rs762204431 Chr.14: 30885472-30885473 on Build GRCh38
rs775897050 Chr.14: 30885571-30885571 on Build GRCh38
rs963288777 Chr.14: 30885430-30885430 on Build GRCh38

View Full Product Details