Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186583776-186584049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB11
Entrez Gene ID
51726
Gene Name
DnaJ heat shock protein family (Hsp40) member B11
Gene Aliases
ABBP-2, ABBP2, DJ9, Dj-9, EDJ, ERdj3, ERj3, ERj3p, PRO1080, UNQ537
UniGene
Hs.317192
Gene Chromosome Location
Chr.3: 186570676-186585800 on Build GRCh38
Transcripts
NM_016306.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3736316 c.790A>G p.I264V Chr.3: 186583914-186583914 on Build GRCh38
4115942 c.832A>G p.T278A Chr.3: 186583956-186583956 on Build GRCh38
4115941 c.821A>G p.E274G Chr.3: 186583945-186583945 on Build GRCh38
3332351 c.838T>C p.L280L Chr.3: 186583962-186583962 on Build GRCh38
7178976 c.761G>A p.R254K Chr.3: 186583885-186583885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940157230 Chr.3: 186583857-186583857 on Build GRCh38
rs567082628 Chr.3: 186583959-186583959 on Build GRCh38
rs766597436 Chr.3: 186583916-186583916 on Build GRCh38
rs769929074 Chr.3: 186584021-186584021 on Build GRCh38
rs939670757 Chr.3: 186583928-186583928 on Build GRCh38
rs376706044 Chr.3: 186583861-186583861 on Build GRCh38
rs748269844 Chr.3: 186584015-186584015 on Build GRCh38
rs376069246 Chr.3: 186583811-186583811 on Build GRCh38
rs748420874 Chr.3: 186583892-186583892 on Build GRCh38
rs371489134 Chr.3: 186583994-186583994 on Build GRCh38
rs938246739 Chr.3: 186583950-186583950 on Build GRCh38
rs537874103 Chr.3: 186583883-186583883 on Build GRCh38
rs745733057 Chr.3: 186583863-186583863 on Build GRCh38
rs980569080 Chr.3: 186583809-186583809 on Build GRCh38
rs6444145 Chr.3: 186583996-186583996 on Build GRCh38
rs141441408 Chr.3: 186583872-186583872 on Build GRCh38
rs758378802 Chr.3: 186584010-186584010 on Build GRCh38
rs780091046 Chr.3: 186584011-186584011 on Build GRCh38
rs770162996 Chr.3: 186583896-186583896 on Build GRCh38
rs367669098 Chr.3: 186583989-186583989 on Build GRCh38
rs773267671 Chr.3: 186583908-186583908 on Build GRCh38
rs370017202 Chr.3: 186583997-186583997 on Build GRCh38
rs752177486 Chr.3: 186583972-186583972 on Build GRCh38
rs8147 Chr.3: 186583914-186583914 on Build GRCh38
rs774983131 Chr.3: 186583918-186583918 on Build GRCh38
rs756334034 Chr.3: 186584023-186584023 on Build GRCh38
rs878950942 Chr.3: 186583879-186583879 on Build GRCh38
rs767057233 Chr.3: 186583938-186583938 on Build GRCh38
rs755568757 Chr.3: 186583986-186583986 on Build GRCh38
rs1055320772 Chr.3: 186583969-186583969 on Build GRCh38
rs760036158 Chr.3: 186583923-186583923 on Build GRCh38

View Full Product Details