Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93347953-93348210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX8C
Entrez Gene ID
341947
Gene Name
cytochrome c oxidase subunit 8C
Gene Aliases
COX8-3
UniGene
Hs.666459
Gene Chromosome Location
Chr.14: 93347191-93348354 on Build GRCh38
Transcripts
NM_182971.2 NM_020818.3 XM_011537027.2 XM_011537028.2

Gene Symbol
UNC79
Entrez Gene ID
57578
Gene Name
unc-79 homolog (C. elegans)
Gene Aliases
KIAA1409
UniGene
Hs.126561
Gene Chromosome Location
Chr.14: 93333219-93710473 on Build GRCh38
Transcripts
NM_182971.2 NM_020818.3 XM_011537027.2 XM_011537028.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1721735 c.127G>A p.E43K Chr.14: 93348028-93348028 on Build GRCh38
3499076 c.173C>T p.P58L Chr.14: 93348074-93348074 on Build GRCh38
7271746 c.206T>C p.F69S Chr.14: 93348107-93348107 on Build GRCh38
239431 c.156delT p.F52fs*5 Chr.14: 93348057-93348057 on Build GRCh38
699041 c.159G>A p.T53T Chr.14: 93348060-93348060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs58621064 Chr.14: 93348147-93348147 on Build GRCh38
rs933101217 Chr.14: 93348011-93348011 on Build GRCh38
rs2455903 Chr.14: 93347978-93347978 on Build GRCh38
rs769596880 Chr.14: 93348158-93348158 on Build GRCh38
rs367709603 Chr.14: 93348121-93348121 on Build GRCh38
rs752359205 Chr.14: 93348027-93348027 on Build GRCh38
rs762575598 Chr.14: 93348038-93348038 on Build GRCh38
rs921647544 Chr.14: 93347979-93347979 on Build GRCh38
rs781686315 Chr.14: 93348065-93348065 on Build GRCh38
rs749514228 Chr.14: 93348116-93348116 on Build GRCh38
rs780349352 Chr.14: 93348104-93348104 on Build GRCh38
rs200469290 Chr.14: 93348019-93348019 on Build GRCh38
rs893750241 Chr.14: 93348085-93348085 on Build GRCh38
rs748273462 Chr.14: 93348131-93348131 on Build GRCh38
rs948046442 Chr.14: 93348106-93348106 on Build GRCh38
rs769623662 Chr.14: 93348166-93348166 on Build GRCh38
rs985698243 Chr.14: 93348003-93348003 on Build GRCh38
rs764995014 Chr.14: 93348024-93348024 on Build GRCh38
rs751080904 Chr.14: 93348052-93348052 on Build GRCh38
rs113042438 Chr.14: 93348073-93348073 on Build GRCh38
rs777042835 Chr.14: 93348174-93348175 on Build GRCh38
rs201893758 Chr.14: 93348041-93348041 on Build GRCh38
rs930054122 Chr.14: 93348043-93348043 on Build GRCh38
rs760634155 Chr.14: 93348006-93348006 on Build GRCh38
rs776529596 Chr.14: 93348017-93348017 on Build GRCh38
rs911477710 Chr.14: 93348009-93348009 on Build GRCh38
rs1003634299 Chr.14: 93348162-93348162 on Build GRCh38
rs774353330 Chr.14: 93348001-93348001 on Build GRCh38
rs918785256 Chr.14: 93348021-93348021 on Build GRCh38
rs145148406 Chr.14: 93348081-93348081 on Build GRCh38
rs773528681 Chr.14: 93348063-93348065 on Build GRCh38
rs778986970 Chr.14: 93348130-93348130 on Build GRCh38
rs759582974 Chr.14: 93348164-93348164 on Build GRCh38
rs770166960 Chr.14: 93348046-93348048 on Build GRCh38
rs371446880 Chr.14: 93348012-93348012 on Build GRCh38
rs757692455 Chr.14: 93348059-93348059 on Build GRCh38
rs776692310 Chr.14: 93348152-93348152 on Build GRCh38

View Full Product Details