Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22165473-22165698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP1
Entrez Gene ID
649
Gene Name
bone morphogenetic protein 1
Gene Aliases
OI13, PCOLC, PCP, PCP2, TLD
UniGene
Hs.1274
Gene Chromosome Location
Chr.8: 22165140-22212326 on Build GRCh38
Transcripts
NM_001199.3 NM_006129.4 NR_033403.1 NR_033404.1 XM_006716386.2 XM_011544617.1 XM_017013738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6431427 c.136C>T p.P46S Chr.8: 22165541-22165541 on Build GRCh38
32237 c.133G>C p.D45H Chr.8: 22165538-22165538 on Build GRCh38
6431425 c.136C>T p.P46S Chr.8: 22165541-22165541 on Build GRCh38
6431426 c.136C>T p.P46S Chr.8: 22165541-22165541 on Build GRCh38
5141587 c.94C>A p.L32M Chr.8: 22165499-22165499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547431045 Chr.8: 22165632-22165632 on Build GRCh38
rs144114975 Chr.8: 22165513-22165513 on Build GRCh38
rs762947562 Chr.8: 22165530-22165530 on Build GRCh38
rs368677695 Chr.8: 22165599-22165599 on Build GRCh38
rs954595323 Chr.8: 22165651-22165651 on Build GRCh38
rs767778995 Chr.8: 22165561-22165561 on Build GRCh38
rs528929027 Chr.8: 22165597-22165597 on Build GRCh38
rs764228182 Chr.8: 22165567-22165567 on Build GRCh38
rs986077867 Chr.8: 22165652-22165652 on Build GRCh38
rs1015688476 Chr.8: 22165622-22165622 on Build GRCh38
rs777310526 Chr.8: 22165603-22165603 on Build GRCh38
rs752974413 Chr.8: 22165562-22165562 on Build GRCh38
rs1006012337 Chr.8: 22165601-22165601 on Build GRCh38
rs772305345 Chr.8: 22165559-22165560 on Build GRCh38
rs1013150486 Chr.8: 22165569-22165569 on Build GRCh38
rs771035694 Chr.8: 22165549-22165549 on Build GRCh38
rs772658585 Chr.8: 22165525-22165525 on Build GRCh38
rs111790440 Chr.8: 22165559-22165559 on Build GRCh38
rs941049889 Chr.8: 22165523-22165523 on Build GRCh38
rs780856935 Chr.8: 22165510-22165510 on Build GRCh38
rs561644531 Chr.8: 22165524-22165524 on Build GRCh38
rs747826181 Chr.8: 22165593-22165593 on Build GRCh38
rs1030606260 Chr.8: 22165638-22165638 on Build GRCh38
rs774235911 Chr.8: 22165550-22165550 on Build GRCh38
rs757759490 Chr.8: 22165575-22165575 on Build GRCh38
rs1039129623 Chr.8: 22165582-22165582 on Build GRCh38
rs760862788 Chr.8: 22165563-22165563 on Build GRCh38
rs747098694 Chr.8: 22165568-22165569 on Build GRCh38
rs990593158 Chr.8: 22165506-22165506 on Build GRCh38
rs746281763 Chr.8: 22165503-22165503 on Build GRCh38
rs780767093 Chr.8: 22165591-22165591 on Build GRCh38
rs895992751 Chr.8: 22165607-22165607 on Build GRCh38
rs759431739 Chr.8: 22165552-22165552 on Build GRCh38
rs1022913854 Chr.8: 22165592-22165592 on Build GRCh38
rs933943218 Chr.8: 22165566-22165566 on Build GRCh38
rs971943478 Chr.8: 22165635-22165635 on Build GRCh38
rs1036680473 Chr.8: 22165542-22165542 on Build GRCh38
rs773383208 Chr.8: 22165561-22165561 on Build GRCh38
rs768249198 Chr.8: 22165504-22165504 on Build GRCh38
rs376619963 Chr.8: 22165574-22165574 on Build GRCh38
rs750747298 Chr.8: 22165583-22165583 on Build GRCh38

View Full Product Details