Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32128599-32128872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBPL
Entrez Gene ID
63943
Gene Name
FK506 binding protein like
Gene Aliases
DIR1, FKBP4, NG7, WISP39
UniGene
Hs.520042
Gene Chromosome Location
Chr.6: 32128707-32130290 on Build GRCh38
Transcripts
NM_022110.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1077824 c.988G>A p.V330I Chr.6: 32128793-32128793 on Build GRCh38
6664343 c.964G>A p.A322T Chr.6: 32128817-32128817 on Build GRCh38
4827712 c.1002G>A p.K334K Chr.6: 32128779-32128779 on Build GRCh38
108650 c.951C>T p.P317P Chr.6: 32128830-32128830 on Build GRCh38
3872918 c.1033C>T p.R345C Chr.6: 32128748-32128748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779007293 Chr.6: 32128732-32128732 on Build GRCh38
rs758373997 Chr.6: 32128697-32128697 on Build GRCh38
rs773168673 Chr.6: 32128823-32128823 on Build GRCh38
rs1051727689 Chr.6: 32128659-32128659 on Build GRCh38
rs781584405 Chr.6: 32128761-32128761 on Build GRCh38
rs762384115 Chr.6: 32128794-32128794 on Build GRCh38
rs61747172 Chr.6: 32128751-32128751 on Build GRCh38
rs566224759 Chr.6: 32128809-32128809 on Build GRCh38
rs768272927 Chr.6: 32128795-32128795 on Build GRCh38
rs746326623 Chr.6: 32128765-32128765 on Build GRCh38
rs866750883 Chr.6: 32128647-32128647 on Build GRCh38
rs767488978 Chr.6: 32128701-32128701 on Build GRCh38
rs771254707 Chr.6: 32128753-32128753 on Build GRCh38
rs761601745 Chr.6: 32128813-32128813 on Build GRCh38
rs1014589960 Chr.6: 32128839-32128839 on Build GRCh38
rs752720310 Chr.6: 32128733-32128733 on Build GRCh38
rs770235355 Chr.6: 32128782-32128782 on Build GRCh38
rs758586156 Chr.6: 32128740-32128740 on Build GRCh38
rs988039370 Chr.6: 32128827-32128827 on Build GRCh38
rs751344069 Chr.6: 32128681-32128681 on Build GRCh38
rs750983954 Chr.6: 32128667-32128667 on Build GRCh38
rs146973385 Chr.6: 32128830-32128830 on Build GRCh38
rs946387958 Chr.6: 32128840-32128840 on Build GRCh38
rs1026549579 Chr.6: 32128650-32128650 on Build GRCh38
rs750358744 Chr.6: 32128714-32128714 on Build GRCh38
rs1006195348 Chr.6: 32128658-32128658 on Build GRCh38
rs539528029 Chr.6: 32128748-32128748 on Build GRCh38
rs368593035 Chr.6: 32128687-32128687 on Build GRCh38
rs147280001 Chr.6: 32128784-32128784 on Build GRCh38
rs764295325 Chr.6: 32128709-32128713 on Build GRCh38
rs760590647 Chr.6: 32128847-32128847 on Build GRCh38
rs767401151 Chr.6: 32128817-32128817 on Build GRCh38
rs914128223 Chr.6: 32128654-32128654 on Build GRCh38
rs751739118 Chr.6: 32128720-32128723 on Build GRCh38
rs993333491 Chr.6: 32128730-32128730 on Build GRCh38
rs756058814 Chr.6: 32128723-32128723 on Build GRCh38
rs970130307 Chr.6: 32128686-32128688 on Build GRCh38
rs933584604 Chr.6: 32128656-32128656 on Build GRCh38
rs913806792 Chr.6: 32128829-32128829 on Build GRCh38

View Full Product Details