Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44786017-44786254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST4
Entrez Gene ID
266675
Gene Name
bestrophin 4
Gene Aliases
VMD2L2
UniGene
Hs.302513
Gene Chromosome Location
Chr.1: 44781836-44792828 on Build GRCh38
Transcripts
NM_153274.2 XM_017001020.1 XM_017001021.1 XM_017001022.1 XM_017001023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
138196 c.518G>A p.S173N Chr.1: 44786192-44786192 on Build GRCh38
3490147 c.633G>A p.L211L Chr.1: 44786077-44786077 on Build GRCh38
6611366 c.573G>T p.L191L Chr.1: 44786137-44786137 on Build GRCh38
7150683 c.567C>T p.T189T Chr.1: 44786143-44786143 on Build GRCh38
6611364 c.587G>A p.R196Q Chr.1: 44786123-44786123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768770759 Chr.1: 44786134-44786134 on Build GRCh38
rs757722278 Chr.1: 44786216-44786216 on Build GRCh38
rs774605499 Chr.1: 44786117-44786117 on Build GRCh38
rs906381528 Chr.1: 44786097-44786097 on Build GRCh38
rs900741062 Chr.1: 44786148-44786148 on Build GRCh38
rs746656859 Chr.1: 44786095-44786095 on Build GRCh38
rs199576518 Chr.1: 44786135-44786135 on Build GRCh38
rs779510830 Chr.1: 44786163-44786163 on Build GRCh38
rs375272773 Chr.1: 44786058-44786058 on Build GRCh38
rs766264466 Chr.1: 44786199-44786199 on Build GRCh38
rs764980649 Chr.1: 44786051-44786051 on Build GRCh38
rs1016270608 Chr.1: 44786200-44786200 on Build GRCh38
rs760870126 Chr.1: 44786130-44786130 on Build GRCh38
rs756426747 Chr.1: 44786088-44786088 on Build GRCh38
rs1043735831 Chr.1: 44786192-44786193 on Build GRCh38
rs772289177 Chr.1: 44786124-44786124 on Build GRCh38
rs952595402 Chr.1: 44786156-44786156 on Build GRCh38
rs761686766 Chr.1: 44786143-44786143 on Build GRCh38
rs754596132 Chr.1: 44786105-44786105 on Build GRCh38
rs368354581 Chr.1: 44786233-44786233 on Build GRCh38
rs771431937 Chr.1: 44786115-44786115 on Build GRCh38
rs201590874 Chr.1: 44786234-44786234 on Build GRCh38
rs750198222 Chr.1: 44786168-44786168 on Build GRCh38
rs780600364 Chr.1: 44786214-44786214 on Build GRCh38
rs779794071 Chr.1: 44786094-44786094 on Build GRCh38
rs868534965 Chr.1: 44786229-44786229 on Build GRCh38
rs890631760 Chr.1: 44786067-44786067 on Build GRCh38
rs376760826 Chr.1: 44786237-44786237 on Build GRCh38
rs762768363 Chr.1: 44786187-44786187 on Build GRCh38
rs765171294 Chr.1: 44786163-44786163 on Build GRCh38
rs201152880 Chr.1: 44786164-44786164 on Build GRCh38
rs142027051 Chr.1: 44786050-44786050 on Build GRCh38
rs749800944 Chr.1: 44786112-44786112 on Build GRCh38
rs758144669 Chr.1: 44786065-44786065 on Build GRCh38
rs145787289 Chr.1: 44786209-44786209 on Build GRCh38
rs141277899 Chr.1: 44786122-44786122 on Build GRCh38

View Full Product Details