Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48177266-48177537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC25A
Entrez Gene ID
993
Gene Name
cell division cycle 25A
Gene Aliases
CDC25A2
UniGene
Hs.437705
Gene Chromosome Location
Chr.3: 48157178-48188658 on Build GRCh38
Transcripts
NM_001789.2 NM_201567.1 XM_006713434.2 XM_006713435.2 XM_011534316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7329970 c.735C>T p.L245L Chr.3: 48177392-48177392 on Build GRCh38
4770480 c.706T>A p.C236S Chr.3: 48177421-48177421 on Build GRCh38
5486362 c.704C>T p.S235L Chr.3: 48177423-48177423 on Build GRCh38
6624149 c.707G>A p.C236Y Chr.3: 48177420-48177420 on Build GRCh38
1753246 c.687T>G p.N229K Chr.3: 48177440-48177440 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs951027610 Chr.3: 48177317-48177317 on Build GRCh38
rs761433140 Chr.3: 48177335-48177335 on Build GRCh38
rs775989648 Chr.3: 48177331-48177331 on Build GRCh38
rs370788532 Chr.3: 48177490-48177490 on Build GRCh38
rs748846112 Chr.3: 48177378-48177380 on Build GRCh38
rs199710455 Chr.3: 48177483-48177483 on Build GRCh38
rs747614606 Chr.3: 48177476-48177476 on Build GRCh38
rs781227114 Chr.3: 48177469-48177469 on Build GRCh38
rs764713762 Chr.3: 48177399-48177399 on Build GRCh38
rs368979184 Chr.3: 48177339-48177339 on Build GRCh38
rs770405298 Chr.3: 48177303-48177303 on Build GRCh38
rs767877083 Chr.3: 48177348-48177348 on Build GRCh38
rs527346675 Chr.3: 48177314-48177314 on Build GRCh38
rs933175747 Chr.3: 48177496-48177496 on Build GRCh38
rs769402476 Chr.3: 48177336-48177336 on Build GRCh38
rs746878109 Chr.3: 48177440-48177440 on Build GRCh38
rs538196915 Chr.3: 48177463-48177463 on Build GRCh38
rs757446929 Chr.3: 48177420-48177420 on Build GRCh38
rs761075879 Chr.3: 48177392-48177392 on Build GRCh38
rs753832415 Chr.3: 48177415-48177415 on Build GRCh38
rs560309143 Chr.3: 48177311-48177311 on Build GRCh38
rs772880657 Chr.3: 48177337-48177337 on Build GRCh38
rs368546115 Chr.3: 48177423-48177423 on Build GRCh38
rs866453474 Chr.3: 48177500-48177500 on Build GRCh38
rs779709401 Chr.3: 48177431-48177431 on Build GRCh38
rs762592987 Chr.3: 48177492-48177492 on Build GRCh38
rs572329670 Chr.3: 48177388-48177388 on Build GRCh38
rs768705877 Chr.3: 48177324-48177324 on Build GRCh38
rs145991453 Chr.3: 48177422-48177422 on Build GRCh38
rs144655393 Chr.3: 48177509-48177509 on Build GRCh38

View Full Product Details