Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40514677-40514950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXO5
Entrez Gene ID
64789
Gene Name
exonuclease 5
Gene Aliases
C1orf176, DEM1, Exo V, hExo5
UniGene
Hs.59584
Gene Chromosome Location
Chr.1: 40508719-40516556 on Build GRCh38
Transcripts
NM_022774.1 XM_005271125.3 XM_005271126.3 XM_005271128.3 XM_011541970.2 XM_017002099.1 XM_017002100.1 XM_017002101.1 XM_017002102.1 XM_017002103.1 XM_017002104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
146484 c.301G>A p.E101K Chr.1: 40514845-40514845 on Build GRCh38
6352251 c.343G>A p.D115N Chr.1: 40514887-40514887 on Build GRCh38
1687517 c.151C>T p.L51F Chr.1: 40514695-40514695 on Build GRCh38
4928454 c.326A>G p.E109G Chr.1: 40514870-40514870 on Build GRCh38
2169259 c.189G>A p.W63* Chr.1: 40514733-40514733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890704813 Chr.1: 40514852-40514852 on Build GRCh38
rs150426595 Chr.1: 40514859-40514859 on Build GRCh38
rs1011293239 Chr.1: 40514898-40514898 on Build GRCh38
rs753947910 Chr.1: 40514920-40514920 on Build GRCh38
rs1048010150 Chr.1: 40514867-40514867 on Build GRCh38
rs965565347 Chr.1: 40514770-40514770 on Build GRCh38
rs369655272 Chr.1: 40514876-40514876 on Build GRCh38
rs376563843 Chr.1: 40514772-40514772 on Build GRCh38
rs757512390 Chr.1: 40514806-40514806 on Build GRCh38
rs918491703 Chr.1: 40514787-40514787 on Build GRCh38
rs768644767 Chr.1: 40514850-40514850 on Build GRCh38
rs764901873 Chr.1: 40514760-40514760 on Build GRCh38
rs747677933 Chr.1: 40514866-40514866 on Build GRCh38
rs774647599 Chr.1: 40514761-40514761 on Build GRCh38
rs201053995 Chr.1: 40514706-40514706 on Build GRCh38
rs750215357 Chr.1: 40514819-40514819 on Build GRCh38
rs746864844 Chr.1: 40514844-40514844 on Build GRCh38
rs775933877 Chr.1: 40514893-40514893 on Build GRCh38
rs373242015 Chr.1: 40514917-40514917 on Build GRCh38
rs200406212 Chr.1: 40514762-40514762 on Build GRCh38
rs764019469 Chr.1: 40514788-40514788 on Build GRCh38
rs760791949 Chr.1: 40514777-40514777 on Build GRCh38
rs562627911 Chr.1: 40514701-40514701 on Build GRCh38
rs911084576 Chr.1: 40514742-40514742 on Build GRCh38
rs753968355 Chr.1: 40514793-40514793 on Build GRCh38
rs780642620 Chr.1: 40514857-40514857 on Build GRCh38
rs746416501 Chr.1: 40514786-40514787 on Build GRCh38
rs761018348 Chr.1: 40514895-40514895 on Build GRCh38
rs199704407 Chr.1: 40514741-40514741 on Build GRCh38
rs758198157 Chr.1: 40514830-40514830 on Build GRCh38
rs772891136 Chr.1: 40514880-40514880 on Build GRCh38
rs1134586 Chr.1: 40514887-40514887 on Build GRCh38
rs764726501 Chr.1: 40514897-40514897 on Build GRCh38
rs761391346 Chr.1: 40514714-40514714 on Build GRCh38
rs759940866 Chr.1: 40514881-40514881 on Build GRCh38
rs1055932311 Chr.1: 40514912-40514912 on Build GRCh38
rs780031828 Chr.1: 40514838-40514838 on Build GRCh38
rs779328840 Chr.1: 40514815-40514815 on Build GRCh38

View Full Product Details