Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 39660918-39661191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT5
Entrez Gene ID
10317
Gene Name
beta-1,3-galactosyltransferase 5
Gene Aliases
B3GalT-V, B3GalTx, B3T5, GLCT5, beta-1,3-GalTase 5, beta-3-Gx-T5, beta3Gal-T5
UniGene
Hs.733638
Gene Chromosome Location
Chr.21: 39612940-39662889 on Build GRCh38
Transcripts
NM_001278650.1 NM_006057.2 NM_033170.2 NM_033171.2 NM_033172.2 XM_017028227.1 XM_017028228.1 XM_017028229.1 XM_017028230.1 XM_017028231.1 XM_017028232.1 XM_017028233.1 XM_017028234.1 XM_017028235.1 XM_017028236.1 XM_017028237.1 XM_017028238.1 XM_017028239.1 XM_017028240.1 XM_017028241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2818413 c.402G>T p.K134N Chr.21: 39660961-39660961 on Build GRCh38
2818415 c.574A>T p.R192W Chr.21: 39661133-39661133 on Build GRCh38
6609244 c.525A>G p.R175R Chr.21: 39661084-39661084 on Build GRCh38
7175118 c.367G>A p.D123N Chr.21: 39660926-39660926 on Build GRCh38
279096 c.449C>T p.A150V Chr.21: 39661008-39661008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530374938 Chr.21: 39661119-39661119 on Build GRCh38
rs146963445 Chr.21: 39660987-39660987 on Build GRCh38
rs772798170 Chr.21: 39660963-39660963 on Build GRCh38
rs776315199 Chr.21: 39660968-39660968 on Build GRCh38
rs754309316 Chr.21: 39661074-39661074 on Build GRCh38
rs779012920 Chr.21: 39661001-39661001 on Build GRCh38
rs778291682 Chr.21: 39660989-39660989 on Build GRCh38
rs759358783 Chr.21: 39661057-39661057 on Build GRCh38
rs371036613 Chr.21: 39660972-39660972 on Build GRCh38
rs544780257 Chr.21: 39661059-39661059 on Build GRCh38
rs867543548 Chr.21: 39661127-39661127 on Build GRCh38
rs534470099 Chr.21: 39660961-39660961 on Build GRCh38
rs374097178 Chr.21: 39661052-39661052 on Build GRCh38
rs368725752 Chr.21: 39660980-39660980 on Build GRCh38
rs913540616 Chr.21: 39661091-39661091 on Build GRCh38
rs375341407 Chr.21: 39661159-39661159 on Build GRCh38
rs150903728 Chr.21: 39661053-39661053 on Build GRCh38
rs769516181 Chr.21: 39661111-39661111 on Build GRCh38
rs734411 Chr.21: 39660990-39660990 on Build GRCh38
rs748112479 Chr.21: 39661108-39661108 on Build GRCh38
rs781050314 Chr.21: 39661104-39661104 on Build GRCh38
rs753240764 Chr.21: 39660985-39660985 on Build GRCh38
rs768724281 Chr.21: 39661102-39661102 on Build GRCh38
rs147073249 Chr.21: 39660999-39660999 on Build GRCh38
rs757960329 Chr.21: 39660974-39660974 on Build GRCh38
rs368422023 Chr.21: 39661118-39661118 on Build GRCh38
rs775691952 Chr.21: 39661006-39661006 on Build GRCh38
rs761527376 Chr.21: 39661034-39661034 on Build GRCh38
rs750726351 Chr.21: 39661090-39661090 on Build GRCh38
rs770598841 Chr.21: 39661150-39661150 on Build GRCh38
rs780385319 Chr.21: 39661092-39661092 on Build GRCh38
rs554528663 Chr.21: 39660969-39660969 on Build GRCh38
rs372607334 Chr.21: 39661005-39661005 on Build GRCh38
rs571754735 Chr.21: 39660957-39660957 on Build GRCh38
rs754034712 Chr.21: 39660983-39660983 on Build GRCh38
rs779308972 Chr.21: 39661087-39661087 on Build GRCh38
rs776567576 Chr.21: 39661029-39661029 on Build GRCh38
rs201449827 Chr.21: 39661035-39661035 on Build GRCh38
rs369712108 Chr.21: 39661044-39661044 on Build GRCh38
rs374024196 Chr.21: 39660973-39660973 on Build GRCh38
rs538924308 Chr.21: 39661009-39661009 on Build GRCh38
rs734412 Chr.21: 39661135-39661135 on Build GRCh38
rs767418363 Chr.21: 39661082-39661082 on Build GRCh38
rs764510727 Chr.21: 39661060-39661060 on Build GRCh38
rs769582300 Chr.21: 39661123-39661124 on Build GRCh38

View Full Product Details