Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19496139-19496380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATAD2A
Entrez Gene ID
54815
Gene Name
GATA zinc finger domain containing 2A
Gene Aliases
p66alpha
UniGene
Hs.118964
Gene Chromosome Location
Chr.19: 19385833-19508932 on Build GRCh38
Transcripts
NM_001300946.1 NM_017660.3 XM_011528105.1 XM_017026901.1 XM_017026902.1 XM_017026903.1 XM_017026904.1 XM_017026905.1 XM_017026906.1 XM_017026907.1 XM_017026908.1 XM_017026909.1 XM_017026910.1 XM_017026911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7105066 c.444C>T p.L148L Chr.19: 19496168-19496168 on Build GRCh38
7105067 c.873C>T p.L291L Chr.19: 19496168-19496168 on Build GRCh38
5933814 c.862C>T p.Q288* Chr.19: 19496157-19496157 on Build GRCh38
7105068 c.873C>T p.L291L Chr.19: 19496168-19496168 on Build GRCh38
5800326 c.920C>G p.P307R Chr.19: 19496215-19496215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367981840 Chr.19: 19496209-19496209 on Build GRCh38
rs906267179 Chr.19: 19496250-19496250 on Build GRCh38
rs917637524 Chr.19: 19496165-19496165 on Build GRCh38
rs768250542 Chr.19: 19496174-19496174 on Build GRCh38
rs756000158 Chr.19: 19496168-19496168 on Build GRCh38
rs967411617 Chr.19: 19496356-19496356 on Build GRCh38
rs964101106 Chr.19: 19496301-19496301 on Build GRCh38
rs777887495 Chr.19: 19496281-19496281 on Build GRCh38
rs754770631 Chr.19: 19496264-19496264 on Build GRCh38
rs756055279 Chr.19: 19496243-19496243 on Build GRCh38
rs779506191 Chr.19: 19496238-19496239 on Build GRCh38
rs748452735 Chr.19: 19496245-19496245 on Build GRCh38
rs758748693 Chr.19: 19496251-19496252 on Build GRCh38
rs1040514025 Chr.19: 19496320-19496320 on Build GRCh38
rs374829526 Chr.19: 19496261-19496261 on Build GRCh38
rs78079701 Chr.19: 19496286-19496286 on Build GRCh38
rs750387323 Chr.19: 19496237-19496237 on Build GRCh38
rs761132140 Chr.19: 19496183-19496183 on Build GRCh38
rs138530563 Chr.19: 19496285-19496285 on Build GRCh38
rs916281264 Chr.19: 19496230-19496230 on Build GRCh38
rs759934826 Chr.19: 19496193-19496193 on Build GRCh38
rs771588933 Chr.19: 19496178-19496178 on Build GRCh38
rs549188396 Chr.19: 19496177-19496177 on Build GRCh38
rs752937530 Chr.19: 19496227-19496227 on Build GRCh38
rs779960713 Chr.19: 19496256-19496257 on Build GRCh38
rs546990310 Chr.19: 19496317-19496317 on Build GRCh38
rs1029714966 Chr.19: 19496342-19496342 on Build GRCh38
rs2288851 Chr.19: 19496182-19496182 on Build GRCh38
rs374965734 Chr.19: 19496172-19496172 on Build GRCh38
rs771291766 Chr.19: 19496184-19496184 on Build GRCh38
rs778621123 Chr.19: 19496175-19496175 on Build GRCh38
rs998129083 Chr.19: 19496313-19496313 on Build GRCh38
rs533125805 Chr.19: 19496294-19496294 on Build GRCh38
rs780124023 Chr.19: 19496257-19496257 on Build GRCh38
rs977875266 Chr.19: 19496206-19496206 on Build GRCh38
rs528154163 Chr.19: 19496187-19496187 on Build GRCh38
rs371903708 Chr.19: 19496231-19496231 on Build GRCh38
rs748958687 Chr.19: 19496173-19496173 on Build GRCh38

View Full Product Details