Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92082251-92082508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN3
Entrez Gene ID
4287
Gene Name
ataxin 3
Gene Aliases
AT3, ATX3, JOS, MJD, MJD1, SCA3
UniGene
Hs.532632
Gene Chromosome Location
Chr.14: 92058552-92106621 on Build GRCh38
Transcripts
NM_001127696.1 NM_001127697.2 NM_001164774.1 NM_001164776.1 NM_001164777.1 NM_001164778.1 NM_001164779.1 NM_001164780.1 NM_001164781.1 NM_001164782.1 NM_004993.5 NM_030660.4 NR_028453.1 NR_028454.1 NR_028455.1 NR_028456.1 NR_028457.1 NR_028458.1 NR_028459.1 NR_028460.1 NR_028461.1 NR_028462.1 NR_028463.1 NR_028464.1 NR_028465.1 NR_028466.1 NR_028467.1 NR_028468.1 NR_028469.1 NR_028470.1 NR_031765.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4858867 c.753G>T p.R251S Chr.14: 92082322-92082322 on Build GRCh38
4410603 c.572A>T p.K191I Chr.14: 92082458-92082458 on Build GRCh38
6476690 c.656C>T p.A219V Chr.14: 92082374-92082374 on Build GRCh38
6476688 c.701C>T p.A234V Chr.14: 92082374-92082374 on Build GRCh38
958846 c.715G>T p.E239* Chr.14: 92082360-92082360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377574592 Chr.14: 92082405-92082405 on Build GRCh38
rs748855843 Chr.14: 92082473-92082473 on Build GRCh38
rs148366775 Chr.14: 92082403-92082403 on Build GRCh38
rs770983214 Chr.14: 92082418-92082418 on Build GRCh38
rs767543973 Chr.14: 92082351-92082351 on Build GRCh38
rs928404702 Chr.14: 92082306-92082306 on Build GRCh38
rs371037244 Chr.14: 92082406-92082406 on Build GRCh38
rs751329134 Chr.14: 92082340-92082340 on Build GRCh38
rs776828308 Chr.14: 92082326-92082326 on Build GRCh38
rs752168522 Chr.14: 92082434-92082434 on Build GRCh38
rs891248894 Chr.14: 92082415-92082415 on Build GRCh38
rs746109327 Chr.14: 92082426-92082426 on Build GRCh38
rs143402326 Chr.14: 92082352-92082352 on Build GRCh38
rs553632954 Chr.14: 92082480-92082480 on Build GRCh38
rs747156311 Chr.14: 92082416-92082416 on Build GRCh38
rs777522387 Chr.14: 92082392-92082394 on Build GRCh38
rs199790369 Chr.14: 92082357-92082357 on Build GRCh38
rs889026927 Chr.14: 92082396-92082396 on Build GRCh38
rs774064667 Chr.14: 92082431-92082431 on Build GRCh38
rs773271553 Chr.14: 92082439-92082439 on Build GRCh38
rs759748483 Chr.14: 92082327-92082327 on Build GRCh38
rs755842617 Chr.14: 92082374-92082382 on Build GRCh38
rs754605940 Chr.14: 92082469-92082469 on Build GRCh38
rs138404351 Chr.14: 92082443-92082443 on Build GRCh38
rs761679000 Chr.14: 92082350-92082350 on Build GRCh38
rs754590483 Chr.14: 92082393-92082393 on Build GRCh38
rs760721207 Chr.14: 92082318-92082318 on Build GRCh38
rs199876727 Chr.14: 92082365-92082365 on Build GRCh38
rs771181207 Chr.14: 92082322-92082322 on Build GRCh38
rs201553525 Chr.14: 92082346-92082346 on Build GRCh38
rs764919608 Chr.14: 92082470-92082470 on Build GRCh38
rs1048755 Chr.14: 92082441-92082441 on Build GRCh38
rs142148800 Chr.14: 92082433-92082433 on Build GRCh38
rs75188275 Chr.14: 92082425-92082425 on Build GRCh38
rs773301187 Chr.14: 92082304-92082304 on Build GRCh38
rs770009219 Chr.14: 92082430-92082430 on Build GRCh38
rs772588044 Chr.14: 92082291-92082291 on Build GRCh38
rs201012097 Chr.14: 92082373-92082373 on Build GRCh38
rs779172677 Chr.14: 92082395-92082395 on Build GRCh38
rs753344410 Chr.14: 92082468-92082468 on Build GRCh38
rs763802729 Chr.14: 92082328-92082328 on Build GRCh38
rs200483057 Chr.14: 92082353-92082353 on Build GRCh38
rs748604134 Chr.14: 92082288-92082288 on Build GRCh38
rs149131253 Chr.14: 92082324-92082324 on Build GRCh38
rs548763388 Chr.14: 92082366-92082366 on Build GRCh38

View Full Product Details