Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74525855-74526110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300LB
Entrez Gene ID
124599
Gene Name
CD300 molecule like family member b
Gene Aliases
CD300b, CLM-7, CLM7, CMRF35-A2, IREM-3, IREM3, TREM-5, TREM5
UniGene
Hs.313343
Gene Chromosome Location
Chr.17: 74521174-74531474 on Build GRCh38
Transcripts
NM_174892.3 XM_005257027.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
984067 c.288G>T p.K96N Chr.17: 74525941-74525941 on Build GRCh38
219305 c.238G>A p.E80K Chr.17: 74525991-74525991 on Build GRCh38
984068 c.227A>G p.K76R Chr.17: 74526002-74526002 on Build GRCh38
252423 c.334C>T p.R112C Chr.17: 74525895-74525895 on Build GRCh38
4391780 c.271C>T p.R91C Chr.17: 74525958-74525958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763963716 Chr.17: 74525902-74525902 on Build GRCh38
rs867594822 Chr.17: 74525931-74525931 on Build GRCh38
rs534302161 Chr.17: 74526039-74526039 on Build GRCh38
rs867137802 Chr.17: 74525920-74525920 on Build GRCh38
rs201350611 Chr.17: 74526029-74526029 on Build GRCh38
rs150773470 Chr.17: 74525957-74525957 on Build GRCh38
rs752893026 Chr.17: 74525958-74525958 on Build GRCh38
rs200754539 Chr.17: 74526041-74526041 on Build GRCh38
rs754025653 Chr.17: 74526059-74526059 on Build GRCh38
rs1037409676 Chr.17: 74525916-74525916 on Build GRCh38
rs771582861 Chr.17: 74526004-74526004 on Build GRCh38
rs537689621 Chr.17: 74525969-74525969 on Build GRCh38
rs777910263 Chr.17: 74526054-74526054 on Build GRCh38
rs376345140 Chr.17: 74526080-74526080 on Build GRCh38
rs201815587 Chr.17: 74525996-74525996 on Build GRCh38
rs112562175 Chr.17: 74525891-74525891 on Build GRCh38
rs114130167 Chr.17: 74526019-74526019 on Build GRCh38
rs1030445762 Chr.17: 74526031-74526031 on Build GRCh38
rs756186631 Chr.17: 74526053-74526053 on Build GRCh38
rs750343010 Chr.17: 74525956-74525956 on Build GRCh38
rs546240009 Chr.17: 74525894-74525894 on Build GRCh38
rs535338624 Chr.17: 74525922-74525922 on Build GRCh38
rs752840212 Chr.17: 74526044-74526044 on Build GRCh38
rs111972938 Chr.17: 74526025-74526025 on Build GRCh38
rs779424173 Chr.17: 74525994-74525994 on Build GRCh38
rs954847095 Chr.17: 74526036-74526036 on Build GRCh38
rs1030861378 Chr.17: 74525983-74525983 on Build GRCh38
rs889719772 Chr.17: 74526048-74526048 on Build GRCh38
rs1043573740 Chr.17: 74526015-74526015 on Build GRCh38
rs757064453 Chr.17: 74525896-74525896 on Build GRCh38
rs548897430 Chr.17: 74525945-74525945 on Build GRCh38
rs147451231 Chr.17: 74526020-74526020 on Build GRCh38
rs141231081 Chr.17: 74525917-74525917 on Build GRCh38
rs199893448 Chr.17: 74526042-74526042 on Build GRCh38
rs199976982 Chr.17: 74525918-74525918 on Build GRCh38
rs764966030 Chr.17: 74526037-74526037 on Build GRCh38
rs756384831 Chr.17: 74525960-74525960 on Build GRCh38
rs528149782 Chr.17: 74525982-74525982 on Build GRCh38
rs568686362 Chr.17: 74525966-74525966 on Build GRCh38
rs759025254 Chr.17: 74525878-74525878 on Build GRCh38
rs199715046 Chr.17: 74525938-74525938 on Build GRCh38
rs761612833 Chr.17: 74526030-74526030 on Build GRCh38
rs139180365 Chr.17: 74525967-74525967 on Build GRCh38
rs199687179 Chr.17: 74525895-74525895 on Build GRCh38

View Full Product Details