Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39532491-39532758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EID2B
Entrez Gene ID
126272
Gene Name
EP300 interacting inhibitor of differentiation 2B
Gene Aliases
EID-2B, EID-3
UniGene
Hs.135181
Gene Chromosome Location
Chr.19: 39531533-39532867 on Build GRCh38
Transcripts
NM_152361.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6843048 c.120T>A p.V40V Chr.19: 39532683-39532683 on Build GRCh38
6843047 c.121C>G p.R41G Chr.19: 39532682-39532682 on Build GRCh38
1681021 c.63G>T p.Q21H Chr.19: 39532740-39532740 on Build GRCh38
1630924 c.139G>C p.E47Q Chr.19: 39532664-39532664 on Build GRCh38
7214165 c.153delC p.E52fs*37 Chr.19: 39532650-39532650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753715110 Chr.19: 39532728-39532728 on Build GRCh38
rs752442157 Chr.19: 39532730-39532730 on Build GRCh38
rs775839575 Chr.19: 39532608-39532608 on Build GRCh38
rs759181726 Chr.19: 39532729-39532729 on Build GRCh38
rs759274357 Chr.19: 39532628-39532628 on Build GRCh38
rs1123301 Chr.19: 39532668-39532668 on Build GRCh38
rs764740279 Chr.19: 39532547-39532547 on Build GRCh38
rs759868599 Chr.19: 39532530-39532530 on Build GRCh38
rs774688840 Chr.19: 39532526-39532526 on Build GRCh38
rs1002893859 Chr.19: 39532570-39532570 on Build GRCh38
rs757988349 Chr.19: 39532552-39532552 on Build GRCh38
rs980495259 Chr.19: 39532618-39532618 on Build GRCh38
rs746116428 Chr.19: 39532605-39532605 on Build GRCh38
rs749902072 Chr.19: 39532548-39532548 on Build GRCh38
rs550180281 Chr.19: 39532641-39532641 on Build GRCh38
rs769033614 Chr.19: 39532610-39532610 on Build GRCh38
rs377527673 Chr.19: 39532611-39532611 on Build GRCh38
rs373058868 Chr.19: 39532704-39532704 on Build GRCh38
rs1031927519 Chr.19: 39532557-39532557 on Build GRCh38
rs919167631 Chr.19: 39532713-39532713 on Build GRCh38
rs770290842 Chr.19: 39532712-39532712 on Build GRCh38
rs746025797 Chr.19: 39532689-39532689 on Build GRCh38
rs145347341 Chr.19: 39532562-39532562 on Build GRCh38
rs200823121 Chr.19: 39532615-39532615 on Build GRCh38
rs761187141 Chr.19: 39532538-39532538 on Build GRCh38
rs750651509 Chr.19: 39532564-39532565 on Build GRCh38
rs757410773 Chr.19: 39532523-39532524 on Build GRCh38
rs752535802 Chr.19: 39532636-39532636 on Build GRCh38
rs754782304 Chr.19: 39532564-39532564 on Build GRCh38
rs542808511 Chr.19: 39532584-39532584 on Build GRCh38
rs917422972 Chr.19: 39532613-39532613 on Build GRCh38
rs748040458 Chr.19: 39532573-39532573 on Build GRCh38
rs765926354 Chr.19: 39532559-39532559 on Build GRCh38
rs868278339 Chr.19: 39532663-39532663 on Build GRCh38
rs749311445 Chr.19: 39532593-39532593 on Build GRCh38
rs778969782 Chr.19: 39532604-39532604 on Build GRCh38
rs747356364 Chr.19: 39532699-39532699 on Build GRCh38
rs1000412867 Chr.19: 39532551-39532551 on Build GRCh38
rs778983825 Chr.19: 39532669-39532669 on Build GRCh38
rs769104709 Chr.19: 39532700-39532700 on Build GRCh38
rs767672229 Chr.19: 39532560-39532561 on Build GRCh38
rs150463695 Chr.19: 39532515-39532515 on Build GRCh38
rs756149965 Chr.19: 39532613-39532614 on Build GRCh38
rs369438240 Chr.19: 39532692-39532692 on Build GRCh38
rs751209565 Chr.19: 39532625-39532625 on Build GRCh38
rs776964290 Chr.19: 39532701-39532701 on Build GRCh38
rs777832923 Chr.19: 39532588-39532588 on Build GRCh38
rs767907651 Chr.19: 39532535-39532535 on Build GRCh38
rs767353677 Chr.19: 39532634-39532634 on Build GRCh38
rs781158249 Chr.19: 39532569-39532569 on Build GRCh38
rs753850231 Chr.19: 39532660-39532660 on Build GRCh38
rs771088143 Chr.19: 39532603-39532603 on Build GRCh38
rs967244812 Chr.19: 39532675-39532675 on Build GRCh38
rs377059957 Chr.19: 39532721-39532721 on Build GRCh38
rs931106433 Chr.19: 39532518-39532518 on Build GRCh38
rs772367081 Chr.19: 39532607-39532607 on Build GRCh38
rs780130396 Chr.19: 39532699-39532700 on Build GRCh38
rs780315608 Chr.19: 39532697-39532697 on Build GRCh38
rs757260648 Chr.19: 39532617-39532617 on Build GRCh38
rs761099135 Chr.19: 39532609-39532609 on Build GRCh38
rs909423503 Chr.19: 39532529-39532529 on Build GRCh38

View Full Product Details