Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55746141-55746366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX4
Entrez Gene ID
54514
Gene Name
DEAD-box helicase 4
Gene Aliases
VASA
UniGene
Hs.223581
Gene Chromosome Location
Chr.5: 55737994-55817146 on Build GRCh38
Transcripts
NM_001142549.1 NM_001166533.1 NM_024415.2 XM_011543495.2 XM_011543497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6428628 c.92G>T p.G31V Chr.5: 55746186-55746186 on Build GRCh38
5749231 c.122C>T p.S41L Chr.5: 55746216-55746216 on Build GRCh38
5403647 c.83G>A p.G28E Chr.5: 55746177-55746177 on Build GRCh38
5975630 c.111T>C p.T37T Chr.5: 55746205-55746205 on Build GRCh38
233926 c.70-9T>G p.? Chr.5: 55746155-55746155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771635664 Chr.5: 55746185-55746185 on Build GRCh38
rs770871592 Chr.5: 55746219-55746219 on Build GRCh38
rs146565989 Chr.5: 55746183-55746183 on Build GRCh38
rs141308424 Chr.5: 55746205-55746205 on Build GRCh38
rs751695480 Chr.5: 55746270-55746270 on Build GRCh38
rs1022581373 Chr.5: 55746318-55746318 on Build GRCh38
rs901368507 Chr.5: 55746287-55746287 on Build GRCh38
rs769316385 Chr.5: 55746236-55746236 on Build GRCh38
rs1012418666 Chr.5: 55746309-55746309 on Build GRCh38
rs776759889 Chr.5: 55746228-55746228 on Build GRCh38
rs199859572 Chr.5: 55746264-55746264 on Build GRCh38
rs376529344 Chr.5: 55746240-55746240 on Build GRCh38
rs745321927 Chr.5: 55746233-55746233 on Build GRCh38
rs959552369 Chr.5: 55746297-55746297 on Build GRCh38
rs761754206 Chr.5: 55746273-55746273 on Build GRCh38
rs777288263 Chr.5: 55746200-55746200 on Build GRCh38
rs778530167 Chr.5: 55746177-55746177 on Build GRCh38
rs992940475 Chr.5: 55746173-55746173 on Build GRCh38
rs200278733 Chr.5: 55746217-55746217 on Build GRCh38
rs777812202 Chr.5: 55746265-55746265 on Build GRCh38
rs897240055 Chr.5: 55746232-55746232 on Build GRCh38
rs868584994 Chr.5: 55746209-55746209 on Build GRCh38
rs775432520 Chr.5: 55746249-55746249 on Build GRCh38
rs73127426 Chr.5: 55746335-55746335 on Build GRCh38
rs998292292 Chr.5: 55746329-55746329 on Build GRCh38
rs147762531 Chr.5: 55746334-55746334 on Build GRCh38
rs759393265 Chr.5: 55746234-55746234 on Build GRCh38

View Full Product Details