Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9760539-9760810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK1
Entrez Gene ID
8536
Gene Name
calcium/calmodulin dependent protein kinase I
Gene Aliases
CAMKI
UniGene
Hs.434875
Gene Chromosome Location
Chr.3: 9757345-9769987 on Build GRCh38
Transcripts
NM_003656.4 XM_005265516.1 XM_005265517.2 XM_017007354.1 NM_016821.2 NM_016826.2 NM_016827.2 NM_016828.2 NM_016829.2 XM_011533760.1 XM_017006494.1 XM_017006496.1 XM_017006497.1 XM_017006498.1 XM_017006499.1

Gene Symbol
OGG1
Entrez Gene ID
4968
Gene Name
8-oxoguanine DNA glycosylase
Gene Aliases
HMMH, HOGG1, MUTM, OGH1
UniGene
Hs.380271
Gene Chromosome Location
Chr.3: 9749944-9788246 on Build GRCh38
Transcripts
NM_003656.4 XM_005265516.1 XM_005265517.2 XM_017007354.1 NM_016821.2 NM_016826.2 NM_016827.2 NM_016828.2 NM_016829.2 XM_011533760.1 XM_017006494.1 XM_017006496.1 XM_017006497.1 XM_017006498.1 XM_017006499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1049185 c.640G>A p.G214S Chr.3: 9760761-9760761 on Build GRCh38
4120775 c.635T>C p.L212P Chr.3: 9760766-9760766 on Build GRCh38
6619157 c.699C>T p.A233A Chr.3: 9760702-9760702 on Build GRCh38
6209226 c.745+3A>T p.? Chr.3: 9760653-9760653 on Build GRCh38
4823871 c.737C>G p.S246C Chr.3: 9760664-9760664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747682193 Chr.3: 9760671-9760671 on Build GRCh38
rs143557505 Chr.3: 9760696-9760696 on Build GRCh38
rs201794302 Chr.3: 9760639-9760639 on Build GRCh38
rs962834816 Chr.3: 9760564-9760564 on Build GRCh38
rs750784176 Chr.3: 9760658-9760658 on Build GRCh38
rs1041670618 Chr.3: 9760738-9760738 on Build GRCh38
rs140837712 Chr.3: 9760663-9760663 on Build GRCh38
rs765649218 Chr.3: 9760653-9760653 on Build GRCh38
rs945881488 Chr.3: 9760699-9760699 on Build GRCh38
rs747165508 Chr.3: 9760701-9760701 on Build GRCh38
rs1044423442 Chr.3: 9760589-9760589 on Build GRCh38
rs747592160 Chr.3: 9760607-9760607 on Build GRCh38
rs777310726 Chr.3: 9760674-9760674 on Build GRCh38
rs143142172 Chr.3: 9760672-9760672 on Build GRCh38
rs779963851 Chr.3: 9760788-9760788 on Build GRCh38
rs918830322 Chr.3: 9760582-9760582 on Build GRCh38
rs947368957 Chr.3: 9760583-9760583 on Build GRCh38
rs986857770 Chr.3: 9760624-9760624 on Build GRCh38
rs767556194 Chr.3: 9760636-9760636 on Build GRCh38
rs147073343 Chr.3: 9760746-9760746 on Build GRCh38
rs972980547 Chr.3: 9760571-9760571 on Build GRCh38
rs200459492 Chr.3: 9760702-9760702 on Build GRCh38
rs760839181 Chr.3: 9760718-9760718 on Build GRCh38
rs377198670 Chr.3: 9760652-9760652 on Build GRCh38
rs753380998 Chr.3: 9760763-9760763 on Build GRCh38
rs752050008 Chr.3: 9760759-9760759 on Build GRCh38
rs746977343 Chr.3: 9760789-9760789 on Build GRCh38
rs762008030 Chr.3: 9760740-9760740 on Build GRCh38
rs369091347 Chr.3: 9760775-9760775 on Build GRCh38
rs770601593 Chr.3: 9760695-9760695 on Build GRCh38
rs55722214 Chr.3: 9760669-9760669 on Build GRCh38
rs141086851 Chr.3: 9760566-9760569 on Build GRCh38
rs749142057 Chr.3: 9760622-9760622 on Build GRCh38
rs748937664 Chr.3: 9760689-9760689 on Build GRCh38
rs138951531 Chr.3: 9760762-9760762 on Build GRCh38
rs770675137 Chr.3: 9760625-9760625 on Build GRCh38
rs756971878 Chr.3: 9760767-9760767 on Build GRCh38
rs775518138 Chr.3: 9760705-9760705 on Build GRCh38
rs188481420 Chr.3: 9760596-9760596 on Build GRCh38
rs774260794 Chr.3: 9760629-9760629 on Build GRCh38
rs372672840 Chr.3: 9760642-9760642 on Build GRCh38
rs755492997 Chr.3: 9760761-9760761 on Build GRCh38
rs769503002 Chr.3: 9760614-9760614 on Build GRCh38
rs760612412 Chr.3: 9760640-9760640 on Build GRCh38
rs766905823 Chr.3: 9760750-9760750 on Build GRCh38
rs759945660 Chr.3: 9760669-9760671 on Build GRCh38
rs768657576 Chr.3: 9760721-9760721 on Build GRCh38
rs553868707 Chr.3: 9760563-9760563 on Build GRCh38
rs907102077 Chr.3: 9760752-9760752 on Build GRCh38
rs759601422 Chr.3: 9760630-9760630 on Build GRCh38
rs371280138 Chr.3: 9760778-9760778 on Build GRCh38
rs955100180 Chr.3: 9760588-9760588 on Build GRCh38
rs531632583 Chr.3: 9760666-9760666 on Build GRCh38
rs772693575 Chr.3: 9760619-9760619 on Build GRCh38
rs753967585 Chr.3: 9760651-9760651 on Build GRCh38
rs910996592 Chr.3: 9760635-9760635 on Build GRCh38
rs765378672 Chr.3: 9760741-9760741 on Build GRCh38
rs1003571829 Chr.3: 9760780-9760780 on Build GRCh38
rs778648261 Chr.3: 9760772-9760772 on Build GRCh38
rs1048619930 Chr.3: 9760753-9760753 on Build GRCh38

View Full Product Details