Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 28023670-28023925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF18A
Entrez Gene ID
81930
Gene Name
kinesin family member 18A
Gene Aliases
MS-KIF18A, PPP1R99
UniGene
Hs.301052
Gene Chromosome Location
Chr.11: 28020616-28108199 on Build GRCh38
Transcripts
NM_031217.3 XM_017018379.1 XM_017018380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
71337 c.2517C>G p.N839K Chr.11: 28023838-28023838 on Build GRCh38
4453130 c.2572A>C p.N858H Chr.11: 28023783-28023783 on Build GRCh38
926155 c.2522C>T p.S841L Chr.11: 28023833-28023833 on Build GRCh38
5838390 c.2533G>C p.D845H Chr.11: 28023822-28023822 on Build GRCh38
926154 c.2543C>A p.S848Y Chr.11: 28023812-28023812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767693064 Chr.11: 28023721-28023721 on Build GRCh38
rs754587319 Chr.11: 28023883-28023883 on Build GRCh38
rs770203259 Chr.11: 28023703-28023703 on Build GRCh38
rs368276152 Chr.11: 28023711-28023711 on Build GRCh38
rs773750645 Chr.11: 28023826-28023826 on Build GRCh38
rs751776880 Chr.11: 28023716-28023716 on Build GRCh38
rs915374424 Chr.11: 28023874-28023874 on Build GRCh38
rs766137624 Chr.11: 28023869-28023869 on Build GRCh38
rs754257705 Chr.11: 28023885-28023885 on Build GRCh38
rs765126083 Chr.11: 28023855-28023855 on Build GRCh38
rs751308160 Chr.11: 28023884-28023885 on Build GRCh38
rs750367086 Chr.11: 28023856-28023856 on Build GRCh38
rs397849132 Chr.11: 28023891-28023891 on Build GRCh38
rs746647574 Chr.11: 28023807-28023807 on Build GRCh38
rs778788771 Chr.11: 28023762-28023762 on Build GRCh38
rs759776183 Chr.11: 28023838-28023838 on Build GRCh38
rs984352634 Chr.11: 28023883-28023884 on Build GRCh38
rs143135596 Chr.11: 28023795-28023795 on Build GRCh38
rs573052783 Chr.11: 28023802-28023802 on Build GRCh38
rs373149170 Chr.11: 28023850-28023850 on Build GRCh38
rs773412995 Chr.11: 28023708-28023708 on Build GRCh38
rs780705673 Chr.11: 28023891-28023891 on Build GRCh38
rs759990590 Chr.11: 28023865-28023865 on Build GRCh38
rs952585171 Chr.11: 28023797-28023797 on Build GRCh38
rs540161804 Chr.11: 28023810-28023810 on Build GRCh38
rs993992882 Chr.11: 28023717-28023717 on Build GRCh38
rs1054661171 Chr.11: 28023776-28023776 on Build GRCh38
rs757220711 Chr.11: 28023752-28023752 on Build GRCh38
rs33937991 Chr.11: 28023713-28023713 on Build GRCh38
rs199512613 Chr.11: 28023816-28023816 on Build GRCh38
rs754101012 Chr.11: 28023883-28023883 on Build GRCh38
rs112609277 Chr.11: 28023748-28023748 on Build GRCh38
rs759756095 Chr.11: 28023719-28023719 on Build GRCh38
rs767735994 Chr.11: 28023848-28023848 on Build GRCh38
rs369809315 Chr.11: 28023833-28023833 on Build GRCh38
rs148846163 Chr.11: 28023851-28023851 on Build GRCh38
rs750232861 Chr.11: 28023785-28023785 on Build GRCh38
rs12273233 Chr.11: 28023872-28023872 on Build GRCh38
rs748687672 Chr.11: 28023819-28023819 on Build GRCh38
rs34088899 Chr.11: 28023884-28023884 on Build GRCh38
rs753830666 Chr.11: 28023747-28023747 on Build GRCh38
rs35440657 Chr.11: 28023820-28023820 on Build GRCh38

View Full Product Details