Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244689146-244689407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DESI2
Entrez Gene ID
51029
Gene Name
desumoylating isopeptidase 2
Gene Aliases
C1orf121, CGI-146, DESI, DESI1, DeSI-2, FAM152A, PNAS-4, PPPDE1
UniGene
Hs.498317
Gene Chromosome Location
Chr.1: 244653048-244709034 on Build GRCh38
Transcripts
NM_001297746.1 NM_016076.4 NR_123735.1 NR_123736.1 XM_011544203.2 XM_011544205.2 XM_011544206.2 XM_011544207.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5793018 c.184G>A p.E62K Chr.1: 244689317-244689317 on Build GRCh38
1320295 c.172G>A p.G58R Chr.1: 244689305-244689305 on Build GRCh38
4470737 c.167C>T p.S56F Chr.1: 244689300-244689300 on Build GRCh38
3804622 c.116-1G>T p.? Chr.1: 244689248-244689248 on Build GRCh38
7236987 c.191G>C p.G64A Chr.1: 244689324-244689324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759987328 Chr.1: 244689300-244689300 on Build GRCh38
rs914985618 Chr.1: 244689355-244689355 on Build GRCh38
rs990449542 Chr.1: 244689183-244689183 on Build GRCh38
rs13375259 Chr.1: 244689283-244689283 on Build GRCh38
rs774710840 Chr.1: 244689276-244689276 on Build GRCh38
rs949154794 Chr.1: 244689198-244689198 on Build GRCh38
rs749032484 Chr.1: 244689229-244689229 on Build GRCh38
rs762100695 Chr.1: 244689381-244689381 on Build GRCh38
rs555860728 Chr.1: 244689200-244689200 on Build GRCh38
rs990606797 Chr.1: 244689217-244689217 on Build GRCh38
rs771130707 Chr.1: 244689274-244689274 on Build GRCh38
rs916248830 Chr.1: 244689191-244689191 on Build GRCh38
rs760723144 Chr.1: 244689332-244689332 on Build GRCh38
rs780804493 Chr.1: 244689199-244689199 on Build GRCh38
rs772596867 Chr.1: 244689308-244689308 on Build GRCh38
rs937643200 Chr.1: 244689179-244689179 on Build GRCh38
rs369225487 Chr.1: 244689380-244689380 on Build GRCh38
rs776015505 Chr.1: 244689319-244689319 on Build GRCh38
rs777275122 Chr.1: 244689228-244689228 on Build GRCh38
rs764293803 Chr.1: 244689376-244689376 on Build GRCh38
rs769392677 Chr.1: 244689203-244689203 on Build GRCh38

View Full Product Details