Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49373945-49374218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNB1
Entrez Gene ID
3745
Gene Name
potassium voltage-gated channel subfamily B member 1
Gene Aliases
DRK1, Kv2.1
UniGene
Hs.633143
Gene Chromosome Location
Chr.20: 49363877-49484033 on Build GRCh38
Transcripts
NM_004975.3 XM_006723784.3 XM_011528799.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3799643 c.1595T>G p.M532R Chr.20: 49373965-49373965 on Build GRCh38
3547569 c.1508C>T p.S503F Chr.20: 49374052-49374052 on Build GRCh38
312149 c.1414A>G p.M472V Chr.20: 49374146-49374146 on Build GRCh38
1713646 c.1406G>A p.G469E Chr.20: 49374154-49374154 on Build GRCh38
3547568 c.1522G>A p.E508K Chr.20: 49374038-49374038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995149478 Chr.20: 49374141-49374141 on Build GRCh38
rs770317185 Chr.20: 49374189-49374189 on Build GRCh38
rs767889610 Chr.20: 49374031-49374031 on Build GRCh38
rs770677266 Chr.20: 49374010-49374010 on Build GRCh38
rs773970060 Chr.20: 49374017-49374017 on Build GRCh38
rs140932636 Chr.20: 49373984-49373984 on Build GRCh38
rs750701951 Chr.20: 49374114-49374114 on Build GRCh38
rs150200332 Chr.20: 49374081-49374081 on Build GRCh38
rs764502655 Chr.20: 49374062-49374062 on Build GRCh38
rs1009356716 Chr.20: 49373972-49373973 on Build GRCh38
rs761247010 Chr.20: 49374054-49374054 on Build GRCh38
rs777560545 Chr.20: 49374173-49374173 on Build GRCh38
rs759934364 Chr.20: 49374029-49374029 on Build GRCh38
rs377030406 Chr.20: 49374009-49374009 on Build GRCh38
rs1040014511 Chr.20: 49374107-49374107 on Build GRCh38
rs565445245 Chr.20: 49374146-49374146 on Build GRCh38
rs773800413 Chr.20: 49374192-49374192 on Build GRCh38
rs775774087 Chr.20: 49374042-49374042 on Build GRCh38
rs115929614 Chr.20: 49374171-49374171 on Build GRCh38
rs368043123 Chr.20: 49374194-49374194 on Build GRCh38
rs748879098 Chr.20: 49374174-49374174 on Build GRCh38
rs761082551 Chr.20: 49374193-49374193 on Build GRCh38
rs185899368 Chr.20: 49373979-49373979 on Build GRCh38
rs868575516 Chr.20: 49374090-49374090 on Build GRCh38
rs758648996 Chr.20: 49374136-49374136 on Build GRCh38
rs372880201 Chr.20: 49374075-49374075 on Build GRCh38
rs944401761 Chr.20: 49374012-49374012 on Build GRCh38
rs754004606 Chr.20: 49374072-49374072 on Build GRCh38
rs748122520 Chr.20: 49373975-49373975 on Build GRCh38
rs145421532 Chr.20: 49374172-49374172 on Build GRCh38
rs138635977 Chr.20: 49374142-49374142 on Build GRCh38

View Full Product Details