Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 36959231-36959504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1328
Entrez Gene ID
57536
Gene Name
KIAA1328
Gene Aliases
-
UniGene
Hs.4045
Gene Chromosome Location
Chr.18: 36828929-37232127 on Build GRCh38
Transcripts
NM_001322327.1 NM_020776.1 NR_136303.1 NR_136304.1 NR_136305.1 XM_005258315.4 XM_005258317.3 XM_005258318.4 XM_011526099.2 XM_011526100.2 XM_017025876.1 XM_017025877.1 XM_017025878.1 XM_017025879.1 XM_017025880.1 XM_017025881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3701362 c.473G>A p.C158Y Chr.18: 36959332-36959332 on Build GRCh38
6082890 c.538G>A p.G180S Chr.18: 36959409-36959409 on Build GRCh38
988262 c.478G>T p.E160* Chr.18: 36959337-36959337 on Build GRCh38
3701363 c.461G>A p.C154Y Chr.18: 36959332-36959332 on Build GRCh38
6524186 c.510G>A p.E170E Chr.18: 36959381-36959381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771407049 Chr.18: 36959441-36959441 on Build GRCh38
rs750087799 Chr.18: 36959360-36959360 on Build GRCh38
rs1046965242 Chr.18: 36959421-36959421 on Build GRCh38
rs767080800 Chr.18: 36959280-36959280 on Build GRCh38
rs771518930 Chr.18: 36959463-36959463 on Build GRCh38
rs774775228 Chr.18: 36959449-36959449 on Build GRCh38
rs370518859 Chr.18: 36959375-36959375 on Build GRCh38
rs769200636 Chr.18: 36959448-36959448 on Build GRCh38
rs776408073 Chr.18: 36959473-36959473 on Build GRCh38
rs370252799 Chr.18: 36959333-36959333 on Build GRCh38
rs201476444 Chr.18: 36959279-36959279 on Build GRCh38
rs771182582 Chr.18: 36959321-36959321 on Build GRCh38
rs772735966 Chr.18: 36959467-36959467 on Build GRCh38
rs753810732 Chr.18: 36959288-36959288 on Build GRCh38
rs760427028 Chr.18: 36959373-36959373 on Build GRCh38
rs776029314 Chr.18: 36959335-36959335 on Build GRCh38
rs906692692 Chr.18: 36959438-36959438 on Build GRCh38
rs373706042 Chr.18: 36959340-36959340 on Build GRCh38
rs376233576 Chr.18: 36959460-36959460 on Build GRCh38
rs747067076 Chr.18: 36959320-36959320 on Build GRCh38
rs376103134 Chr.18: 36959356-36959356 on Build GRCh38
rs760267195 Chr.18: 36959468-36959468 on Build GRCh38
rs757331417 Chr.18: 36959405-36959405 on Build GRCh38
rs187694949 Chr.18: 36959272-36959272 on Build GRCh38
rs371979302 Chr.18: 36959258-36959258 on Build GRCh38
rs368414258 Chr.18: 36959435-36959435 on Build GRCh38
rs754935368 Chr.18: 36959303-36959303 on Build GRCh38
rs562836633 Chr.18: 36959389-36959389 on Build GRCh38
rs763656854 Chr.18: 36959277-36959277 on Build GRCh38
rs200144238 Chr.18: 36959416-36959416 on Build GRCh38
rs776188979 Chr.18: 36959270-36959270 on Build GRCh38
rs759310342 Chr.18: 36959478-36959478 on Build GRCh38
rs770162685 Chr.18: 36959430-36959430 on Build GRCh38
rs759531539 Chr.18: 36959294-36959295 on Build GRCh38
rs749804365 Chr.18: 36959400-36959400 on Build GRCh38
rs976968931 Chr.18: 36959290-36959290 on Build GRCh38
rs539479647 Chr.18: 36959348-36959348 on Build GRCh38
rs746048716 Chr.18: 36959429-36959429 on Build GRCh38
rs898057513 Chr.18: 36959409-36959409 on Build GRCh38
rs777855600 Chr.18: 36959313-36959313 on Build GRCh38
rs368978402 Chr.18: 36959391-36959391 on Build GRCh38
rs372357724 Chr.18: 36959447-36959447 on Build GRCh38
rs761294720 Chr.18: 36959349-36959349 on Build GRCh38
rs779940738 Chr.18: 36959287-36959287 on Build GRCh38
rs368481127 Chr.18: 36959471-36959471 on Build GRCh38
rs778786038 Chr.18: 36959397-36959397 on Build GRCh38
rs781329139 Chr.18: 36959323-36959323 on Build GRCh38
rs763504156 Chr.18: 36959339-36959339 on Build GRCh38
rs981727489 Chr.18: 36959301-36959301 on Build GRCh38
rs747669293 Chr.18: 36959459-36959459 on Build GRCh38
rs773842715 Chr.18: 36959346-36959346 on Build GRCh38
rs770322485 Chr.18: 36959334-36959334 on Build GRCh38
rs755957773 Chr.18: 36959281-36959281 on Build GRCh38

View Full Product Details