Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62308253-62308480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRM1
Entrez Gene ID
11047
Gene Name
adhesion regulating molecule 1
Gene Aliases
ARM-1, ARM1, GP110
UniGene
Hs.90107
Gene Chromosome Location
Chr.20: 62302056-62308862 on Build GRCh38
Transcripts
NM_001281437.1 NM_001281438.1 NM_007002.3 NM_175573.2 XM_005260257.1 XM_011528503.1 XM_017027602.1 XM_017027603.1 XM_017027604.1 XM_017027605.1 XM_017027606.1 XM_017027607.1 XM_017027608.1 XM_017027609.1 XM_017027610.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1196110 c.1046C>T p.S349L Chr.20: 62308399-62308399 on Build GRCh38
4656339 c.1117G>A p.D373N Chr.20: 62308470-62308470 on Build GRCh38
5031837 c.1117+1G>A p.? Chr.20: 62308471-62308471 on Build GRCh38
4137109 c.1061C>A p.P354H Chr.20: 62308414-62308414 on Build GRCh38
6432542 c.1103C>T p.A368V Chr.20: 62308456-62308456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935122651 Chr.20: 62308378-62308378 on Build GRCh38
rs369269114 Chr.20: 62308331-62308331 on Build GRCh38
rs977962780 Chr.20: 62308286-62308286 on Build GRCh38
rs973476573 Chr.20: 62308296-62308296 on Build GRCh38
rs529740931 Chr.20: 62308292-62308292 on Build GRCh38
rs963101455 Chr.20: 62308294-62308294 on Build GRCh38
rs745601273 Chr.20: 62308339-62308339 on Build GRCh38
rs772022918 Chr.20: 62308342-62308342 on Build GRCh38
rs1030463150 Chr.20: 62308277-62308277 on Build GRCh38
rs765985268 Chr.20: 62308431-62308431 on Build GRCh38
rs541680101 Chr.20: 62308430-62308430 on Build GRCh38
rs1033632690 Chr.20: 62308457-62308457 on Build GRCh38
rs182655445 Chr.20: 62308349-62308349 on Build GRCh38
rs887221021 Chr.20: 62308368-62308368 on Build GRCh38
rs11544168 Chr.20: 62308446-62308446 on Build GRCh38
rs200235059 Chr.20: 62308453-62308453 on Build GRCh38
rs754630756 Chr.20: 62308318-62308319 on Build GRCh38
rs775394635 Chr.20: 62308346-62308346 on Build GRCh38
rs748887628 Chr.20: 62308327-62308327 on Build GRCh38
rs767776596 Chr.20: 62308289-62308289 on Build GRCh38
rs114373359 Chr.20: 62308319-62308319 on Build GRCh38
rs150640020 Chr.20: 62308386-62308386 on Build GRCh38
rs547524374 Chr.20: 62308274-62308274 on Build GRCh38
rs753158224 Chr.20: 62308423-62308423 on Build GRCh38
rs369524429 Chr.20: 62308400-62308400 on Build GRCh38
rs767840220 Chr.20: 62308406-62308406 on Build GRCh38
rs934035049 Chr.20: 62308340-62308340 on Build GRCh38
rs764148509 Chr.20: 62308375-62308375 on Build GRCh38
rs867704098 Chr.20: 62308441-62308441 on Build GRCh38
rs748679926 Chr.20: 62308445-62308445 on Build GRCh38
rs572816311 Chr.20: 62308344-62308344 on Build GRCh38
rs1131931 Chr.20: 62308462-62308462 on Build GRCh38
rs190413329 Chr.20: 62308287-62308287 on Build GRCh38
rs756841363 Chr.20: 62308461-62308461 on Build GRCh38
rs760877885 Chr.20: 62308330-62308330 on Build GRCh38
rs773158870 Chr.20: 62308385-62308385 on Build GRCh38
rs766527585 Chr.20: 62308395-62308395 on Build GRCh38

View Full Product Details