Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63239119-63239350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC7
Entrez Gene ID
79444
Gene Name
baculoviral IAP repeat containing 7
Gene Aliases
KIAP, LIVIN, ML-IAP, MLIAP, RNF50
UniGene
Hs.256126
Gene Chromosome Location
Chr.20: 63235883-63240507 on Build GRCh38
Transcripts
NM_022161.3 NM_139317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2931499 c.598G>T p.E200* Chr.20: 63239182-63239182 on Build GRCh38
4912473 c.643G>A p.E215K Chr.20: 63239227-63239227 on Build GRCh38
7221030 c.624C>A p.V208V Chr.20: 63239208-63239208 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772478876 Chr.20: 63239199-63239202 on Build GRCh38
rs765257076 Chr.20: 63239325-63239325 on Build GRCh38
rs778215156 Chr.20: 63239247-63239248 on Build GRCh38
rs758964512 Chr.20: 63239219-63239219 on Build GRCh38
rs560298175 Chr.20: 63239224-63239224 on Build GRCh38
rs781384192 Chr.20: 63239176-63239176 on Build GRCh38
rs905695905 Chr.20: 63239143-63239143 on Build GRCh38
rs775098582 Chr.20: 63239291-63239291 on Build GRCh38
rs1040161389 Chr.20: 63239293-63239293 on Build GRCh38
rs776545168 Chr.20: 63239310-63239310 on Build GRCh38
rs373997317 Chr.20: 63239147-63239147 on Build GRCh38
rs200528640 Chr.20: 63239329-63239329 on Build GRCh38
rs771535832 Chr.20: 63239283-63239283 on Build GRCh38
rs771286092 Chr.20: 63239208-63239208 on Build GRCh38
rs756265998 Chr.20: 63239278-63239278 on Build GRCh38
rs775260228 Chr.20: 63239229-63239229 on Build GRCh38
rs768540333 Chr.20: 63239300-63239302 on Build GRCh38
rs901495544 Chr.20: 63239294-63239294 on Build GRCh38
rs776898586 Chr.20: 63239267-63239282 on Build GRCh38
rs770171399 Chr.20: 63239183-63239183 on Build GRCh38
rs760352384 Chr.20: 63239238-63239238 on Build GRCh38
rs193196289 Chr.20: 63239330-63239330 on Build GRCh38
rs564528998 Chr.20: 63239304-63239304 on Build GRCh38
rs869203806 Chr.20: 63239254-63239255 on Build GRCh38
rs928614092 Chr.20: 63239195-63239195 on Build GRCh38
rs377658110 Chr.20: 63239211-63239211 on Build GRCh38
rs761821260 Chr.20: 63239251-63239251 on Build GRCh38
rs367617870 Chr.20: 63239285-63239285 on Build GRCh38
rs3215649 Chr.20: 63239253-63239254 on Build GRCh38
rs1075556 Chr.20: 63239282-63239282 on Build GRCh38
rs757380953 Chr.20: 63239244-63239244 on Build GRCh38
rs11908302 Chr.20: 63239194-63239194 on Build GRCh38
rs768132359 Chr.20: 63239307-63239307 on Build GRCh38
rs771892125 Chr.20: 63239228-63239228 on Build GRCh38
rs780184137 Chr.20: 63239145-63239145 on Build GRCh38
rs761504955 Chr.20: 63239311-63239311 on Build GRCh38
rs914849161 Chr.20: 63239234-63239234 on Build GRCh38
rs985604368 Chr.20: 63239319-63239319 on Build GRCh38
rs954369067 Chr.20: 63239317-63239317 on Build GRCh38
rs765068567 Chr.20: 63239253-63239253 on Build GRCh38
rs201147952 Chr.20: 63239267-63239267 on Build GRCh38
rs752950376 Chr.20: 63239276-63239276 on Build GRCh38
rs4637179 Chr.20: 63239260-63239260 on Build GRCh38
rs748728178 Chr.20: 63239157-63239158 on Build GRCh38
rs781203230 Chr.20: 63239271-63239271 on Build GRCh38
rs397840121 Chr.20: 63239256-63239257 on Build GRCh38
rs764010820 Chr.20: 63239243-63239243 on Build GRCh38
rs771730459 Chr.20: 63239290-63239290 on Build GRCh38
rs529195260 Chr.20: 63239172-63239172 on Build GRCh38
rs869270953 Chr.20: 63239257-63239258 on Build GRCh38
rs745419633 Chr.20: 63239287-63239287 on Build GRCh38
rs746077865 Chr.20: 63239293-63239296 on Build GRCh38
rs751533808 Chr.20: 63239165-63239165 on Build GRCh38
rs149161695 Chr.20: 63239201-63239201 on Build GRCh38

View Full Product Details