Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185083019-185083246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf70
Entrez Gene ID
285382
Gene Name
chromosome 3 open reading frame 70
Gene Aliases
-
UniGene
Hs.380048
Gene Chromosome Location
Chr.3: 185078050-185153014 on Build GRCh38
Transcripts
NM_001025266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6164080 c.538C>A p.H180N Chr.3: 185083222-185083222 on Build GRCh38
3669116 c.622G>A p.E208K Chr.3: 185083138-185083138 on Build GRCh38
5864065 c.701C>T p.S234F Chr.3: 185083059-185083059 on Build GRCh38
5883789 c.562G>C p.E188Q Chr.3: 185083198-185083198 on Build GRCh38
5699886 c.625G>A p.E209K Chr.3: 185083135-185083135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778071741 Chr.3: 185083061-185083061 on Build GRCh38
rs771250343 Chr.3: 185083155-185083155 on Build GRCh38
rs761501886 Chr.3: 185083182-185083182 on Build GRCh38
rs755723232 Chr.3: 185083132-185083134 on Build GRCh38
rs910937584 Chr.3: 185083131-185083131 on Build GRCh38
rs771483286 Chr.3: 185083069-185083069 on Build GRCh38
rs746170383 Chr.3: 185083162-185083162 on Build GRCh38
rs989092978 Chr.3: 185083071-185083071 on Build GRCh38
rs142152683 Chr.3: 185083056-185083056 on Build GRCh38
rs750196410 Chr.3: 185083140-185083140 on Build GRCh38
rs750259038 Chr.3: 185083199-185083199 on Build GRCh38
rs533213003 Chr.3: 185083150-185083150 on Build GRCh38
rs772665963 Chr.3: 185083163-185083163 on Build GRCh38
rs749591169 Chr.3: 185083154-185083154 on Build GRCh38
rs200872353 Chr.3: 185083147-185083147 on Build GRCh38
rs761448354 Chr.3: 185083112-185083112 on Build GRCh38
rs543744858 Chr.3: 185083080-185083080 on Build GRCh38
rs201783027 Chr.3: 185083053-185083053 on Build GRCh38
rs201641850 Chr.3: 185083048-185083048 on Build GRCh38
rs777542727 Chr.3: 185083148-185083148 on Build GRCh38
rs200917972 Chr.3: 185083178-185083178 on Build GRCh38
rs768220474 Chr.3: 185083176-185083176 on Build GRCh38
rs530327049 Chr.3: 185083169-185083169 on Build GRCh38
rs753433215 Chr.3: 185083103-185083103 on Build GRCh38
rs764820079 Chr.3: 185083197-185083197 on Build GRCh38
rs774855344 Chr.3: 185083073-185083073 on Build GRCh38
rs934213706 Chr.3: 185083123-185083123 on Build GRCh38
rs142754403 Chr.3: 185083134-185083134 on Build GRCh38
rs1049932206 Chr.3: 185083128-185083128 on Build GRCh38
rs777984697 Chr.3: 185083153-185083153 on Build GRCh38
rs781381496 Chr.3: 185083055-185083055 on Build GRCh38
rs746217001 Chr.3: 185083165-185083168 on Build GRCh38
rs779811600 Chr.3: 185083148-185083148 on Build GRCh38
rs752352893 Chr.3: 185083092-185083093 on Build GRCh38
rs762583972 Chr.3: 185083202-185083202 on Build GRCh38
rs770082686 Chr.3: 185083059-185083059 on Build GRCh38
rs146317967 Chr.3: 185083078-185083078 on Build GRCh38
rs775795637 Chr.3: 185083168-185083168 on Build GRCh38
rs913363179 Chr.3: 185083164-185083164 on Build GRCh38
rs994031475 Chr.3: 185083201-185083201 on Build GRCh38
rs139649690 Chr.3: 185083079-185083079 on Build GRCh38
rs760247597 Chr.3: 185083171-185083171 on Build GRCh38
rs760133055 Chr.3: 185083075-185083075 on Build GRCh38
rs779472832 Chr.3: 185083157-185083157 on Build GRCh38
rs756242736 Chr.3: 185083179-185083179 on Build GRCh38
rs756872479 Chr.3: 185083052-185083052 on Build GRCh38
rs1056348337 Chr.3: 185083214-185083214 on Build GRCh38
rs763742130 Chr.3: 185083092-185083092 on Build GRCh38

View Full Product Details