Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18008304-18008553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC2
Entrez Gene ID
27106
Gene Name
arrestin domain containing 2
Gene Aliases
CLONE24945, PP2703
UniGene
Hs.515249
Gene Chromosome Location
Chr.19: 18001132-18014102 on Build GRCh38
Transcripts
NM_001025604.2 NM_001286826.1 NM_015683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7241708 c.61G>C p.E21Q Chr.19: 18008371-18008371 on Build GRCh38
6605222 c.91G>A p.A31T Chr.19: 18008401-18008401 on Build GRCh38
711154 c.225C>T p.Y75Y Chr.19: 18008535-18008535 on Build GRCh38
6575846 c.181G>A p.E61K Chr.19: 18008491-18008491 on Build GRCh38
6905975 c.238G>A p.E80K Chr.19: 18008548-18008548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202031500 Chr.19: 18008504-18008504 on Build GRCh38
rs769492824 Chr.19: 18008457-18008495 on Build GRCh38
rs766387572 Chr.19: 18008381-18008381 on Build GRCh38
rs746877988 Chr.19: 18008466-18008466 on Build GRCh38
rs376508733 Chr.19: 18008525-18008525 on Build GRCh38
rs763867466 Chr.19: 18008508-18008508 on Build GRCh38
rs920695107 Chr.19: 18008495-18008495 on Build GRCh38
rs771782546 Chr.19: 18008374-18008374 on Build GRCh38
rs776694565 Chr.19: 18008388-18008388 on Build GRCh38
rs2100606 Chr.19: 18008325-18008325 on Build GRCh38
rs200613532 Chr.19: 18008397-18008397 on Build GRCh38
rs948320135 Chr.19: 18008527-18008527 on Build GRCh38
rs200200467 Chr.19: 18008363-18008363 on Build GRCh38
rs775184392 Chr.19: 18008505-18008505 on Build GRCh38
rs758847405 Chr.19: 18008352-18008352 on Build GRCh38
rs147126372 Chr.19: 18008355-18008355 on Build GRCh38
rs199505328 Chr.19: 18008407-18008407 on Build GRCh38
rs924058337 Chr.19: 18008502-18008502 on Build GRCh38
rs138687493 Chr.19: 18008349-18008349 on Build GRCh38
rs752008104 Chr.19: 18008422-18008422 on Build GRCh38
rs1002788056 Chr.19: 18008520-18008520 on Build GRCh38
rs1057094096 Chr.19: 18008531-18008531 on Build GRCh38
rs751170141 Chr.19: 18008515-18008515 on Build GRCh38
rs774412489 Chr.19: 18008461-18008461 on Build GRCh38
rs927732305 Chr.19: 18008411-18008411 on Build GRCh38
rs940888899 Chr.19: 18008492-18008492 on Build GRCh38
rs542203111 Chr.19: 18008377-18008377 on Build GRCh38
rs763626910 Chr.19: 18008421-18008421 on Build GRCh38
rs922139670 Chr.19: 18008375-18008375 on Build GRCh38
rs767714554 Chr.19: 18008523-18008523 on Build GRCh38
rs749542607 Chr.19: 18008444-18008444 on Build GRCh38
rs988164490 Chr.19: 18008430-18008430 on Build GRCh38
rs756196800 Chr.19: 18008438-18008438 on Build GRCh38
rs150198820 Chr.19: 18008331-18008331 on Build GRCh38
rs770566707 Chr.19: 18008468-18008468 on Build GRCh38
rs746304456 Chr.19: 18008434-18008434 on Build GRCh38
rs369803137 Chr.19: 18008364-18008364 on Build GRCh38
rs773020251 Chr.19: 18008376-18008376 on Build GRCh38
rs747506436 Chr.19: 18008357-18008357 on Build GRCh38
rs780055385 Chr.19: 18008440-18008440 on Build GRCh38
rs758085197 Chr.19: 18008408-18008408 on Build GRCh38
rs1037739848 Chr.19: 18008493-18008493 on Build GRCh38
rs1028945021 Chr.19: 18008456-18008457 on Build GRCh38
rs759124187 Chr.19: 18008392-18008392 on Build GRCh38
rs768961650 Chr.19: 18008453-18008453 on Build GRCh38
rs749450651 Chr.19: 18008373-18008373 on Build GRCh38
rs775073484 Chr.19: 18008460-18008469 on Build GRCh38
rs148923549 Chr.19: 18008350-18008350 on Build GRCh38
rs553414947 Chr.19: 18008351-18008351 on Build GRCh38
rs769856978 Chr.19: 18008370-18008370 on Build GRCh38
rs370715491 Chr.19: 18008535-18008535 on Build GRCh38
rs762351432 Chr.19: 18008506-18008506 on Build GRCh38
rs757552484 Chr.19: 18008426-18008426 on Build GRCh38
rs776724952 Chr.19: 18008485-18008485 on Build GRCh38
rs781757887 Chr.19: 18008432-18008432 on Build GRCh38
rs954993144 Chr.19: 18008391-18008391 on Build GRCh38
rs759499109 Chr.19: 18008487-18008487 on Build GRCh38
rs781531642 Chr.19: 18008358-18008358 on Build GRCh38

View Full Product Details