Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38110328-38110553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASH2L
Entrez Gene ID
9070
Gene Name
ASH2 like histone lysine methyltransferase complex subunit
Gene Aliases
ASH2, ASH2L1, ASH2L2, Bre2
UniGene
Hs.521530
Gene Chromosome Location
Chr.8: 38104965-38140080 on Build GRCh38
Transcripts
NM_001105214.2 NM_001261832.1 NM_001282272.1 NM_004674.4 XM_005273682.1 XM_005273683.1 XM_006716412.1 XM_006716413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3648805 c.474C>T p.F158F Chr.8: 38110451-38110451 on Build GRCh38
486418 c.402-1G>T p.? Chr.8: 38110378-38110378 on Build GRCh38
1099533 c.415T>C p.F139L Chr.8: 38110392-38110392 on Build GRCh38
3899933 c.479G>A p.R160Q Chr.8: 38110456-38110456 on Build GRCh38
1099534 c.436C>A p.H146N Chr.8: 38110413-38110413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372439679 Chr.8: 38110357-38110357 on Build GRCh38
rs374597283 Chr.8: 38110380-38110380 on Build GRCh38
rs775164877 Chr.8: 38110455-38110455 on Build GRCh38
rs199636518 Chr.8: 38110473-38110473 on Build GRCh38
rs375173227 Chr.8: 38110484-38110484 on Build GRCh38
rs774953379 Chr.8: 38110399-38110399 on Build GRCh38
rs745597253 Chr.8: 38110422-38110422 on Build GRCh38
rs755037193 Chr.8: 38110386-38110386 on Build GRCh38
rs371079556 Chr.8: 38110433-38110433 on Build GRCh38
rs368450890 Chr.8: 38110389-38110389 on Build GRCh38
rs761373235 Chr.8: 38110499-38110499 on Build GRCh38
rs3763555 Chr.8: 38110521-38110521 on Build GRCh38
rs779048897 Chr.8: 38110421-38110421 on Build GRCh38
rs111788457 Chr.8: 38110485-38110485 on Build GRCh38
rs923420341 Chr.8: 38110522-38110522 on Build GRCh38
rs771994471 Chr.8: 38110444-38110444 on Build GRCh38
rs780257596 Chr.8: 38110369-38110369 on Build GRCh38
rs932060877 Chr.8: 38110524-38110524 on Build GRCh38
rs371377585 Chr.8: 38110373-38110373 on Build GRCh38
rs776004262 Chr.8: 38110488-38110488 on Build GRCh38
rs770986602 Chr.8: 38110420-38110420 on Build GRCh38
rs1034004127 Chr.8: 38110356-38110356 on Build GRCh38
rs201749267 Chr.8: 38110505-38110505 on Build GRCh38
rs986122528 Chr.8: 38110530-38110530 on Build GRCh38
rs768399253 Chr.8: 38110486-38110486 on Build GRCh38

View Full Product Details