Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77869645-77869894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAMDC
Entrez Gene ID
28971
Gene Name
adipogenesis associated Mth938 domain containing
Gene Aliases
C11orf67, CK067, PTD015
UniGene
Hs.745040
Gene Chromosome Location
Chr.11: 77821114-77901281 on Build GRCh38
Transcripts
NM_001316957.1 NM_001316958.1 NM_001316960.1 NM_001316961.1 NM_001316962.1 NM_024684.3 NR_133637.1 XM_011544968.2 XM_011544969.2 XM_017017620.1 XM_017017923.1

Gene Symbol
RSF1
Entrez Gene ID
51773
Gene Name
remodeling and spacing factor 1
Gene Aliases
HBXAP, RSF-1, XAP8, p325
UniGene
Hs.420229
Gene Chromosome Location
Chr.11: 77660009-77872232 on Build GRCh38
Transcripts
NM_001316957.1 NM_001316958.1 NM_001316960.1 NM_001316961.1 NM_001316962.1 NM_024684.3 NR_133637.1 XM_011544968.2 XM_011544969.2 XM_017017620.1 XM_017017923.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
339301 c.139C>G p.P47A Chr.11: 77869728-77869728 on Build GRCh38
932212 c.189G>T p.Q63H Chr.11: 77869778-77869778 on Build GRCh38
3398146 c.142G>A p.G48S Chr.11: 77869731-77869731 on Build GRCh38
932211 c.148C>T p.Q50* Chr.11: 77869737-77869737 on Build GRCh38
5961011 c.137C>T p.S46F Chr.11: 77869726-77869726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755674502 Chr.11: 77869680-77869680 on Build GRCh38
rs765873253 Chr.11: 77869813-77869813 on Build GRCh38
rs377312233 Chr.11: 77869795-77869795 on Build GRCh38
rs753464737 Chr.11: 77869716-77869716 on Build GRCh38
rs1054149966 Chr.11: 77869697-77869697 on Build GRCh38
rs373656193 Chr.11: 77869835-77869835 on Build GRCh38
rs879187770 Chr.11: 77869683-77869683 on Build GRCh38
rs764767925 Chr.11: 77869830-77869830 on Build GRCh38
rs375182802 Chr.11: 77869758-77869758 on Build GRCh38
rs371553919 Chr.11: 77869825-77869825 on Build GRCh38
rs758721614 Chr.11: 77869721-77869721 on Build GRCh38
rs892792898 Chr.11: 77869718-77869718 on Build GRCh38
rs773227986 Chr.11: 77869801-77869801 on Build GRCh38
rs542396592 Chr.11: 77869861-77869861 on Build GRCh38
rs769851697 Chr.11: 77869770-77869770 on Build GRCh38
rs577881120 Chr.11: 77869712-77869712 on Build GRCh38
rs760266528 Chr.11: 77869809-77869809 on Build GRCh38
rs1019525775 Chr.11: 77869811-77869811 on Build GRCh38
rs563492746 Chr.11: 77869743-77869743 on Build GRCh38
rs775574982 Chr.11: 77869771-77869771 on Build GRCh38
rs749128498 Chr.11: 77869746-77869746 on Build GRCh38
rs140924044 Chr.11: 77869817-77869817 on Build GRCh38
rs754713270 Chr.11: 77869818-77869818 on Build GRCh38
rs751991125 Chr.11: 77869832-77869832 on Build GRCh38
rs749990904 Chr.11: 77869672-77869672 on Build GRCh38
rs186614523 Chr.11: 77869794-77869794 on Build GRCh38
rs746003091 Chr.11: 77869757-77869757 on Build GRCh38
rs937251200 Chr.11: 77869675-77869675 on Build GRCh38
rs758170308 Chr.11: 77869777-77869778 on Build GRCh38
rs767621885 Chr.11: 77869799-77869799 on Build GRCh38
rs201315751 Chr.11: 77869731-77869731 on Build GRCh38
rs779734082 Chr.11: 77869846-77869847 on Build GRCh38
rs749485820 Chr.11: 77869772-77869772 on Build GRCh38
rs768358907 Chr.11: 77869789-77869789 on Build GRCh38
rs35039902 Chr.11: 77869735-77869735 on Build GRCh38
rs778154745 Chr.11: 77869729-77869729 on Build GRCh38
rs373999180 Chr.11: 77869783-77869783 on Build GRCh38

View Full Product Details