Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 91958874-91959109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
UniGene
Hs.500526
Gene Chromosome Location
Chr.10: 91923769-92030998 on Build GRCh38
Transcripts
NM_003972.2 XM_011540326.1 XM_011540327.2 XM_011540328.2 XM_011540329.2 XM_017016877.1 XM_017016878.1 XM_017016879.1 XM_017016880.1 XM_017016881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3441499 c.938T>C p.L313P Chr.10: 91959102-91959102 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs564869497 Chr.10: 91958928-91958928 on Build GRCh38
rs2275546 Chr.10: 91959028-91959028 on Build GRCh38
rs376827845 Chr.10: 91959040-91959040 on Build GRCh38
rs1036836880 Chr.10: 91958986-91958986 on Build GRCh38
rs570637047 Chr.10: 91958968-91958968 on Build GRCh38
rs532122620 Chr.10: 91958994-91958994 on Build GRCh38
rs1043726925 Chr.10: 91958942-91958942 on Build GRCh38
rs80066036 Chr.10: 91958999-91958999 on Build GRCh38
rs189548960 Chr.10: 91958971-91958971 on Build GRCh38
rs932029040 Chr.10: 91958932-91958932 on Build GRCh38
rs111742597 Chr.10: 91958980-91958980 on Build GRCh38
rs567869060 Chr.10: 91959011-91959011 on Build GRCh38
rs755786703 Chr.10: 91959036-91959036 on Build GRCh38
rs536411732 Chr.10: 91959016-91959016 on Build GRCh38
rs894806357 Chr.10: 91959031-91959031 on Build GRCh38
rs369850383 Chr.10: 91959057-91959057 on Build GRCh38
rs758870215 Chr.10: 91958963-91958964 on Build GRCh38
rs767114896 Chr.10: 91959027-91959027 on Build GRCh38
rs547129263 Chr.10: 91959009-91959009 on Build GRCh38

View Full Product Details