Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52109761-52110034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L10
Entrez Gene ID
10017
Gene Name
BCL2 like 10
Gene Aliases
BCL-B, Boo, Diva, bcl2-L-10
UniGene
Hs.283672
Gene Chromosome Location
Chr.15: 52109263-52112775 on Build GRCh38
Transcripts
NM_001306168.1 NM_020396.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6487005 c.547C>A p.L183M Chr.15: 52109916-52109916 on Build GRCh38
349051 c.510C>T p.F170F Chr.15: 52109953-52109953 on Build GRCh38
2217256 c.523C>T p.P175S Chr.15: 52109940-52109940 on Build GRCh38
3361448 c.568T>C p.C190R Chr.15: 52109895-52109895 on Build GRCh38
6354090 c.614G>A p.*205* Chr.15: 52109849-52109849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747107320 Chr.15: 52109791-52109791 on Build GRCh38
rs757185584 Chr.15: 52109787-52109787 on Build GRCh38
rs767106587 Chr.15: 52109835-52109835 on Build GRCh38
rs200046035 Chr.15: 52109881-52109881 on Build GRCh38
rs777799758 Chr.15: 52109880-52109880 on Build GRCh38
rs796718339 Chr.15: 52109991-52109992 on Build GRCh38
rs370603489 Chr.15: 52109978-52109978 on Build GRCh38
rs774947628 Chr.15: 52109828-52109828 on Build GRCh38
rs932487312 Chr.15: 52109812-52109812 on Build GRCh38
rs778069285 Chr.15: 52109794-52109805 on Build GRCh38
rs540534800 Chr.15: 52109868-52109868 on Build GRCh38
rs752344133 Chr.15: 52109850-52109850 on Build GRCh38
rs528231878 Chr.15: 52109862-52109862 on Build GRCh38
rs374324889 Chr.15: 52109960-52109960 on Build GRCh38
rs760040170 Chr.15: 52109919-52109919 on Build GRCh38
rs779410273 Chr.15: 52109985-52109985 on Build GRCh38
rs770058262 Chr.15: 52109874-52109874 on Build GRCh38
rs373991834 Chr.15: 52109980-52109980 on Build GRCh38
rs2231296 Chr.15: 52109805-52109805 on Build GRCh38
rs1002786894 Chr.15: 52109947-52109947 on Build GRCh38
rs201823962 Chr.15: 52110001-52110001 on Build GRCh38
rs770110645 Chr.15: 52109809-52109809 on Build GRCh38
rs755777950 Chr.15: 52109854-52109854 on Build GRCh38
rs775062648 Chr.15: 52109911-52109911 on Build GRCh38
rs772598672 Chr.15: 52109991-52109992 on Build GRCh38
rs767006509 Chr.15: 52109823-52109823 on Build GRCh38
rs763824623 Chr.15: 52109938-52109938 on Build GRCh38
rs142737063 Chr.15: 52109944-52109944 on Build GRCh38
rs967817102 Chr.15: 52109797-52109797 on Build GRCh38
rs778759102 Chr.15: 52109788-52109788 on Build GRCh38
rs564303916 Chr.15: 52109858-52109858 on Build GRCh38
rs200554368 Chr.15: 52109829-52109829 on Build GRCh38
rs763342059 Chr.15: 52109820-52109820 on Build GRCh38
rs774697388 Chr.15: 52109978-52109978 on Build GRCh38
rs536602654 Chr.15: 52109912-52109912 on Build GRCh38
rs771539743 Chr.15: 52109908-52109908 on Build GRCh38
rs939912912 Chr.15: 52109839-52109839 on Build GRCh38
rs750415498 Chr.15: 52109959-52109959 on Build GRCh38
rs896743045 Chr.15: 52109965-52109965 on Build GRCh38
rs756991240 Chr.15: 52109859-52109859 on Build GRCh38
rs776870688 Chr.15: 52109997-52109997 on Build GRCh38
rs763866209 Chr.15: 52109856-52109856 on Build GRCh38
rs1035170077 Chr.15: 52109948-52109948 on Build GRCh38
rs760100568 Chr.15: 52109896-52109896 on Build GRCh38
rs921143698 Chr.15: 52109808-52109808 on Build GRCh38
rs749242140 Chr.15: 52109786-52109786 on Build GRCh38
rs182161605 Chr.15: 52109883-52109883 on Build GRCh38
rs772639138 Chr.15: 52109991-52109991 on Build GRCh38
rs746412551 Chr.15: 52109978-52109979 on Build GRCh38
rs764992406 Chr.15: 52109950-52109950 on Build GRCh38
rs748378214 Chr.15: 52109806-52109806 on Build GRCh38
rs746444415 Chr.15: 52109986-52109986 on Build GRCh38
rs749549892 Chr.15: 52109913-52109913 on Build GRCh38
rs748381671 Chr.15: 52109870-52109870 on Build GRCh38
rs755097018 Chr.15: 52109867-52109867 on Build GRCh38
rs775039470 Chr.15: 52109885-52109885 on Build GRCh38
rs2231295 Chr.15: 52109813-52109813 on Build GRCh38
rs771394935 Chr.15: 52109965-52109977 on Build GRCh38
rs771473120 Chr.15: 52109982-52109982 on Build GRCh38

View Full Product Details