Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 37391558-37391817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACACA
Entrez Gene ID
31
Gene Name
acetyl-CoA carboxylase alpha
Gene Aliases
ACAC, ACACAD, ACC, ACC1, ACCA
UniGene
Hs.160556
Gene Chromosome Location
Chr.17: 37084992-37406822 on Build GRCh38
Transcripts
NM_198834.2 NM_198839.2 XM_006721853.1 XM_011524701.1 XM_017024553.1 XM_017024554.1 NM_001321399.1 NM_173625.4 XM_011524647.2 XM_011524648.2 XM_011524649.2

Gene Symbol
C17orf78
Entrez Gene ID
284099
Gene Name
chromosome 17 open reading frame 78
Gene Aliases
-
UniGene
Hs.439154
Gene Chromosome Location
Chr.17: 37375710-37392708 on Build GRCh38
Transcripts
NM_198834.2 NM_198839.2 XM_006721853.1 XM_011524701.1 XM_017024553.1 XM_017024554.1 NM_001321399.1 NM_173625.4 XM_011524647.2 XM_011524648.2 XM_011524649.2

SNP Detail

SNP ID Chromosome Location
rs571831923 Chr.17: 37391763-37391763 on Build GRCh38
rs377287082 Chr.17: 37391694-37391694 on Build GRCh38
rs766956579 Chr.17: 37391714-37391714 on Build GRCh38
rs574017441 Chr.17: 37391782-37391782 on Build GRCh38
rs748196404 Chr.17: 37391618-37391618 on Build GRCh38
rs987009260 Chr.17: 37391787-37391787 on Build GRCh38
rs759426583 Chr.17: 37391646-37391646 on Build GRCh38
rs1040683959 Chr.17: 37391789-37391789 on Build GRCh38
rs750899305 Chr.17: 37391728-37391728 on Build GRCh38
rs772062671 Chr.17: 37391621-37391621 on Build GRCh38
rs775244197 Chr.17: 37391657-37391657 on Build GRCh38
rs763731731 Chr.17: 37391680-37391680 on Build GRCh38
rs754238564 Chr.17: 37391766-37391766 on Build GRCh38
rs761326844 Chr.17: 37391683-37391683 on Build GRCh38
rs113066483 Chr.17: 37391755-37391755 on Build GRCh38
rs751176142 Chr.17: 37391682-37391682 on Build GRCh38
rs762795312 Chr.17: 37391678-37391678 on Build GRCh38
rs772306089 Chr.17: 37391779-37391779 on Build GRCh38
rs772160816 Chr.17: 37391612-37391613 on Build GRCh38
rs544163830 Chr.17: 37391613-37391613 on Build GRCh38
rs921591119 Chr.17: 37391740-37391740 on Build GRCh38
rs768461363 Chr.17: 37391610-37391610 on Build GRCh38
rs769764779 Chr.17: 37391648-37391648 on Build GRCh38
rs780593895 Chr.17: 37391602-37391602 on Build GRCh38
rs776584328 Chr.17: 37391643-37391643 on Build GRCh38
rs756655256 Chr.17: 37391743-37391743 on Build GRCh38

View Full Product Details