Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46397756-46397981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMBRA1
Entrez Gene ID
55626
Gene Name
autophagy and beclin 1 regulator 1
Gene Aliases
DCAF3, WDR94
UniGene
Hs.654644
Gene Chromosome Location
Chr.11: 46396412-46594069 on Build GRCh38
Transcripts
NM_001267782.1 NM_001267783.1 NM_001300731.1 NM_017749.3 XM_005253009.3 XM_005253011.3 XM_005253014.3 XM_006718259.2 XM_006718260.2 XM_017018007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1980467 c.3549C>T p.I1183I Chr.11: 46397798-46397798 on Build GRCh38
4191795 c.3405T>C p.G1135G Chr.11: 46397942-46397942 on Build GRCh38
5469607 c.3419A>G p.Y1140C Chr.11: 46397928-46397928 on Build GRCh38
4661343 c.3534C>T p.F1178F Chr.11: 46397813-46397813 on Build GRCh38
189529 c.3311C>T p.T1104M Chr.11: 46397766-46397766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755064472 Chr.11: 46397809-46397809 on Build GRCh38
rs975568389 Chr.11: 46397876-46397876 on Build GRCh38
rs767129001 Chr.11: 46397873-46397873 on Build GRCh38
rs773546892 Chr.11: 46397819-46397819 on Build GRCh38
rs755262492 Chr.11: 46397937-46397937 on Build GRCh38
rs375792318 Chr.11: 46397806-46397806 on Build GRCh38
rs539900531 Chr.11: 46397952-46397952 on Build GRCh38
rs757098489 Chr.11: 46397906-46397906 on Build GRCh38
rs868081487 Chr.11: 46397871-46397871 on Build GRCh38
rs753714405 Chr.11: 46397801-46397801 on Build GRCh38
rs751812076 Chr.11: 46397929-46397929 on Build GRCh38
rs576645378 Chr.11: 46397828-46397828 on Build GRCh38
rs147448508 Chr.11: 46397874-46397874 on Build GRCh38
rs773707115 Chr.11: 46397777-46397777 on Build GRCh38
rs760754455 Chr.11: 46397813-46397813 on Build GRCh38
rs368208385 Chr.11: 46397880-46397880 on Build GRCh38
rs771327564 Chr.11: 46397956-46397956 on Build GRCh38
rs763966082 Chr.11: 46397800-46397800 on Build GRCh38
rs750734087 Chr.11: 46397821-46397821 on Build GRCh38
rs201354368 Chr.11: 46397779-46397779 on Build GRCh38
rs139896964 Chr.11: 46397907-46397907 on Build GRCh38
rs371938280 Chr.11: 46397954-46397954 on Build GRCh38
rs200433904 Chr.11: 46397879-46397879 on Build GRCh38
rs774062481 Chr.11: 46397852-46397852 on Build GRCh38
rs767500993 Chr.11: 46397786-46397786 on Build GRCh38
rs145428746 Chr.11: 46397943-46397943 on Build GRCh38
rs148784670 Chr.11: 46397860-46397860 on Build GRCh38
rs770437654 Chr.11: 46397851-46397851 on Build GRCh38
rs1019878291 Chr.11: 46397858-46397858 on Build GRCh38
rs774642351 Chr.11: 46397957-46397957 on Build GRCh38
rs573180590 Chr.11: 46397798-46397798 on Build GRCh38
rs749900257 Chr.11: 46397798-46397800 on Build GRCh38
rs770347233 Chr.11: 46397818-46397818 on Build GRCh38
rs756586927 Chr.11: 46397946-46397946 on Build GRCh38
rs747012025 Chr.11: 46397808-46397808 on Build GRCh38
rs752683177 Chr.11: 46397790-46397790 on Build GRCh38
rs12295608 Chr.11: 46397861-46397861 on Build GRCh38
rs748652627 Chr.11: 46397816-46397816 on Build GRCh38

View Full Product Details