Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74794161-74794422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTC
Entrez Gene ID
685
Gene Name
betacellulin
Gene Aliases
-
UniGene
Hs.591704
Gene Chromosome Location
Chr.4: 74744759-74794718 on Build GRCh38
Transcripts
NM_001316963.1 NM_001729.3 XM_011532211.1 XM_017008568.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
448126 c.19T>G p.C7G Chr.4: 74794307-74794307 on Build GRCh38
6408218 c.56T>G p.L19R Chr.4: 74794270-74794270 on Build GRCh38
6316446 c.41C>A p.P14Q Chr.4: 74794285-74794285 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs553149527 Chr.4: 74794400-74794400 on Build GRCh38
rs899510640 Chr.4: 74794311-74794311 on Build GRCh38
rs759420286 Chr.4: 74794273-74794273 on Build GRCh38
rs776376746 Chr.4: 74794257-74794257 on Build GRCh38
rs774422158 Chr.4: 74794243-74794247 on Build GRCh38
rs960801939 Chr.4: 74794365-74794365 on Build GRCh38
rs746559703 Chr.4: 74794352-74794352 on Build GRCh38
rs755899826 Chr.4: 74794378-74794378 on Build GRCh38
rs116326617 Chr.4: 74794401-74794401 on Build GRCh38
rs115997569 Chr.4: 74794339-74794339 on Build GRCh38
rs764206086 Chr.4: 74794276-74794276 on Build GRCh38
rs559814573 Chr.4: 74794321-74794321 on Build GRCh38
rs544207492 Chr.4: 74794266-74794266 on Build GRCh38
rs866772616 Chr.4: 74794323-74794323 on Build GRCh38
rs767658564 Chr.4: 74794298-74794298 on Build GRCh38
rs761958860 Chr.4: 74794283-74794283 on Build GRCh38
rs540409136 Chr.4: 74794344-74794344 on Build GRCh38
rs759171115 Chr.4: 74794329-74794329 on Build GRCh38
rs190304442 Chr.4: 74794211-74794211 on Build GRCh38
rs896623015 Chr.4: 74794309-74794309 on Build GRCh38
rs759475370 Chr.4: 74794253-74794253 on Build GRCh38
rs747778808 Chr.4: 74794331-74794331 on Build GRCh38
rs754139857 Chr.4: 74794317-74794317 on Build GRCh38
rs770883015 Chr.4: 74794239-74794239 on Build GRCh38
rs1032640934 Chr.4: 74794341-74794341 on Build GRCh38
rs752227206 Chr.4: 74794314-74794314 on Build GRCh38
rs974742195 Chr.4: 74794204-74794204 on Build GRCh38
rs28549760 Chr.4: 74794307-74794307 on Build GRCh38
rs1010174059 Chr.4: 74794313-74794313 on Build GRCh38
rs750565213 Chr.4: 74794305-74794305 on Build GRCh38
rs919189440 Chr.4: 74794244-74794244 on Build GRCh38
rs985851555 Chr.4: 74794372-74794374 on Build GRCh38
rs185950914 Chr.4: 74794186-74794186 on Build GRCh38
rs541428993 Chr.4: 74794319-74794319 on Build GRCh38
rs574312284 Chr.4: 74794310-74794310 on Build GRCh38
rs751742169 Chr.4: 74794280-74794280 on Build GRCh38

View Full Product Details