Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4747458-4747723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf71
Entrez Gene ID
146562
Gene Name
chromosome 16 open reading frame 71
Gene Aliases
-
UniGene
Hs.602738
Gene Chromosome Location
Chr.16: 4734288-4749396 on Build GRCh38
Transcripts
NM_139170.2 XM_005255144.3 XM_017022975.1 XM_017022976.1 XM_011522453.2

Gene Symbol
ZNF500
Entrez Gene ID
26048
Gene Name
zinc finger protein 500
Gene Aliases
ZKSCAN18, ZSCAN50
UniGene
Hs.513316
Gene Chromosome Location
Chr.16: 4745961-4767219 on Build GRCh38
Transcripts
NM_139170.2 XM_005255144.3 XM_017022975.1 XM_017022976.1 XM_011522453.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6381110 c.1456C>G p.Q486E Chr.16: 4747518-4747518 on Build GRCh38
3888556 c.1478C>T p.P493L Chr.16: 4747540-4747540 on Build GRCh38
1579648 c.1500delG p.D502fs*12 Chr.16: 4747562-4747562 on Build GRCh38
3888555 c.1454G>A p.G485E Chr.16: 4747516-4747516 on Build GRCh38
970929 c.1481G>T p.R494I Chr.16: 4747543-4747543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924805393 Chr.16: 4747681-4747681 on Build GRCh38
rs746316584 Chr.16: 4747546-4747546 on Build GRCh38
rs764136544 Chr.16: 4747524-4747524 on Build GRCh38
rs572416705 Chr.16: 4747650-4747650 on Build GRCh38
rs763493455 Chr.16: 4747667-4747667 on Build GRCh38
rs751429084 Chr.16: 4747588-4747597 on Build GRCh38
rs1015981924 Chr.16: 4747613-4747613 on Build GRCh38
rs753740224 Chr.16: 4747549-4747549 on Build GRCh38
rs780448390 Chr.16: 4747622-4747622 on Build GRCh38
rs943343341 Chr.16: 4747503-4747503 on Build GRCh38
rs1014950377 Chr.16: 4747482-4747482 on Build GRCh38
rs370146169 Chr.16: 4747541-4747541 on Build GRCh38
rs758896042 Chr.16: 4747614-4747614 on Build GRCh38
rs955282421 Chr.16: 4747533-4747533 on Build GRCh38
rs777500019 Chr.16: 4747633-4747633 on Build GRCh38
rs558793296 Chr.16: 4747635-4747635 on Build GRCh38
rs769468323 Chr.16: 4747564-4747564 on Build GRCh38
rs200704525 Chr.16: 4747597-4747597 on Build GRCh38
rs922512626 Chr.16: 4747532-4747532 on Build GRCh38
rs897869140 Chr.16: 4747693-4747693 on Build GRCh38
rs372559816 Chr.16: 4747548-4747548 on Build GRCh38
rs764410000 Chr.16: 4747593-4747593 on Build GRCh38
rs1006318737 Chr.16: 4747671-4747671 on Build GRCh38
rs760848582 Chr.16: 4747588-4747588 on Build GRCh38
rs761465767 Chr.16: 4747508-4747508 on Build GRCh38
rs765238034 Chr.16: 4747599-4747599 on Build GRCh38
rs914638422 Chr.16: 4747699-4747699 on Build GRCh38
rs368970244 Chr.16: 4747661-4747661 on Build GRCh38
rs368403834 Chr.16: 4747529-4747529 on Build GRCh38
rs911097402 Chr.16: 4747552-4747552 on Build GRCh38
rs990639938 Chr.16: 4747689-4747689 on Build GRCh38
rs745856757 Chr.16: 4747649-4747649 on Build GRCh38
rs118001246 Chr.16: 4747481-4747481 on Build GRCh38
rs765362637 Chr.16: 4747663-4747663 on Build GRCh38
rs767807614 Chr.16: 4747520-4747520 on Build GRCh38
rs776916380 Chr.16: 4747659-4747659 on Build GRCh38
rs770569697 Chr.16: 4747638-4747638 on Build GRCh38
rs747365334 Chr.16: 4747630-4747630 on Build GRCh38
rs752865697 Chr.16: 4747523-4747523 on Build GRCh38
rs773993049 Chr.16: 4747644-4747644 on Build GRCh38
rs772381788 Chr.16: 4747480-4747480 on Build GRCh38
rs770512545 Chr.16: 4747578-4747578 on Build GRCh38
rs766096540 Chr.16: 4747580-4747580 on Build GRCh38
rs768279112 Chr.16: 4747653-4747653 on Build GRCh38
rs574712819 Chr.16: 4747639-4747639 on Build GRCh38
rs760541942 Chr.16: 4747652-4747652 on Build GRCh38
rs201318907 Chr.16: 4747534-4747534 on Build GRCh38
rs780270762 Chr.16: 4747582-4747582 on Build GRCh38
rs762845674 Chr.16: 4747516-4747516 on Build GRCh38
rs545222104 Chr.16: 4747567-4747567 on Build GRCh38
rs750998553 Chr.16: 4747609-4747609 on Build GRCh38
rs780395407 Chr.16: 4747557-4747557 on Build GRCh38
rs759479050 Chr.16: 4747580-4747580 on Build GRCh38
rs772768039 Chr.16: 4747513-4747513 on Build GRCh38
rs750677403 Chr.16: 4747550-4747550 on Build GRCh38
rs185132079 Chr.16: 4747684-4747684 on Build GRCh38
rs775341138 Chr.16: 4747584-4747584 on Build GRCh38
rs766937674 Chr.16: 4747669-4747669 on Build GRCh38
rs747816676 Chr.16: 4747558-4747558 on Build GRCh38
rs113245451 Chr.16: 4747664-4747664 on Build GRCh38
rs202150290 Chr.16: 4747517-4747517 on Build GRCh38
rs753925662 Chr.16: 4747527-4747527 on Build GRCh38
rs756203330 Chr.16: 4747569-4747569 on Build GRCh38
rs997931699 Chr.16: 4747696-4747696 on Build GRCh38
rs772634343 Chr.16: 4747566-4747566 on Build GRCh38
rs192580420 Chr.16: 4747518-4747518 on Build GRCh38
rs755255043 Chr.16: 4747632-4747632 on Build GRCh38
rs774438684 Chr.16: 4747579-4747579 on Build GRCh38
rs751958212 Chr.16: 4747677-4747677 on Build GRCh38
rs976083630 Chr.16: 4747531-4747531 on Build GRCh38

View Full Product Details