Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8073638-8073863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX12B
Entrez Gene ID
242
Gene Name
arachidonate 12-lipoxygenase, 12R type
Gene Aliases
12R-LOX, ARCI2
UniGene
Hs.136574
Gene Chromosome Location
Chr.17: 8072636-8087703 on Build GRCh38
Transcripts
NM_001139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598660 c.1727C>T p.A576V Chr.17: 8073685-8073685 on Build GRCh38
985924 c.1729A>G p.K577E Chr.17: 8073683-8073683 on Build GRCh38
985925 c.1695A>G p.R565R Chr.17: 8073717-8073717 on Build GRCh38
5787949 c.1677C>A p.T559T Chr.17: 8073735-8073735 on Build GRCh38
1303517 c.1748C>A p.T583K Chr.17: 8073664-8073664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571659729 Chr.17: 8073814-8073814 on Build GRCh38
rs929558561 Chr.17: 8073774-8073774 on Build GRCh38
rs201543289 Chr.17: 8073764-8073764 on Build GRCh38
rs779544044 Chr.17: 8073799-8073799 on Build GRCh38
rs1005950528 Chr.17: 8073775-8073775 on Build GRCh38
rs767715541 Chr.17: 8073768-8073768 on Build GRCh38
rs1050108042 Chr.17: 8073713-8073713 on Build GRCh38
rs78234608 Chr.17: 8073809-8073809 on Build GRCh38
rs746911380 Chr.17: 8073801-8073801 on Build GRCh38
rs12937410 Chr.17: 8073787-8073787 on Build GRCh38
rs538142063 Chr.17: 8073792-8073792 on Build GRCh38
rs533761499 Chr.17: 8073725-8073725 on Build GRCh38
rs756718354 Chr.17: 8073770-8073770 on Build GRCh38
rs745577308 Chr.17: 8073793-8073793 on Build GRCh38
rs759687516 Chr.17: 8073767-8073767 on Build GRCh38
rs200687206 Chr.17: 8073757-8073757 on Build GRCh38
rs757224692 Chr.17: 8073719-8073719 on Build GRCh38
rs970728898 Chr.17: 8073834-8073834 on Build GRCh38
rs748849529 Chr.17: 8073663-8073663 on Build GRCh38
rs1017374508 Chr.17: 8073746-8073746 on Build GRCh38
rs753184042 Chr.17: 8073769-8073769 on Build GRCh38
rs754207025 Chr.17: 8073784-8073784 on Build GRCh38
rs768605200 Chr.17: 8073804-8073804 on Build GRCh38
rs137853024 Chr.17: 8073678-8073678 on Build GRCh38
rs751773510 Chr.17: 8073766-8073766 on Build GRCh38
rs987598968 Chr.17: 8073740-8073740 on Build GRCh38
rs927924569 Chr.17: 8073806-8073806 on Build GRCh38
rs777947596 Chr.17: 8073783-8073783 on Build GRCh38
rs200345380 Chr.17: 8073756-8073756 on Build GRCh38
rs149842738 Chr.17: 8073837-8073837 on Build GRCh38
rs771514376 Chr.17: 8073797-8073797 on Build GRCh38
rs768375728 Chr.17: 8073735-8073735 on Build GRCh38
rs933001113 Chr.17: 8073677-8073677 on Build GRCh38
rs773921965 Chr.17: 8073694-8073694 on Build GRCh38
rs370446978 Chr.17: 8073723-8073723 on Build GRCh38
rs745351859 Chr.17: 8073708-8073708 on Build GRCh38
rs771635328 Chr.17: 8073714-8073714 on Build GRCh38

View Full Product Details