Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 86875904-86876165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMPR1A
Entrez Gene ID
657
Gene Name
bone morphogenetic protein receptor type 1A
Gene Aliases
10q23del, ACVRLK3, ALK3, CD292, SKR5
UniGene
Hs.524477
Gene Chromosome Location
Chr.10: 86755786-86927969 on Build GRCh38
Transcripts
NM_004329.2 XM_011540103.2 XM_011540104.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6915133 c.3G>T p.M1I Chr.10: 86876021-86876021 on Build GRCh38
3752019 c.4C>A p.P2T Chr.10: 86876022-86876022 on Build GRCh38
3415307 c.67+2T>C p.? Chr.10: 86876087-86876087 on Build GRCh38
6915842 c.4C>T p.P2S Chr.10: 86876022-86876022 on Build GRCh38
3807874 c.11T>A p.L4Q Chr.10: 86876029-86876029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528738896 Chr.10: 86875999-86875999 on Build GRCh38
rs747437716 Chr.10: 86876083-86876083 on Build GRCh38
rs200115604 Chr.10: 86876055-86876055 on Build GRCh38
rs376891757 Chr.10: 86875995-86875995 on Build GRCh38
rs116346662 Chr.10: 86875994-86875994 on Build GRCh38
rs869312758 Chr.10: 86876021-86876021 on Build GRCh38
rs759014147 Chr.10: 86876077-86876077 on Build GRCh38
rs766269417 Chr.10: 86876044-86876044 on Build GRCh38
rs143248687 Chr.10: 86876023-86876023 on Build GRCh38
rs1047677696 Chr.10: 86876013-86876013 on Build GRCh38
rs73351703 Chr.10: 86876131-86876131 on Build GRCh38
rs770909714 Chr.10: 86876121-86876121 on Build GRCh38
rs754752449 Chr.10: 86876048-86876048 on Build GRCh38
rs151235720 Chr.10: 86876065-86876065 on Build GRCh38
rs772744158 Chr.10: 86876116-86876116 on Build GRCh38
rs745920240 Chr.10: 86876071-86876071 on Build GRCh38
rs786203157 Chr.10: 86876019-86876019 on Build GRCh38
rs780704079 Chr.10: 86876078-86876078 on Build GRCh38
rs751434732 Chr.10: 86876113-86876114 on Build GRCh38
rs748682297 Chr.10: 86875997-86875997 on Build GRCh38
rs754015069 Chr.10: 86876056-86876056 on Build GRCh38
rs202004922 Chr.10: 86876134-86876134 on Build GRCh38
rs757299584 Chr.10: 86875976-86875976 on Build GRCh38
rs746411710 Chr.10: 86875992-86875992 on Build GRCh38
rs993642453 Chr.10: 86876038-86876038 on Build GRCh38
rs786204152 Chr.10: 86876084-86876084 on Build GRCh38
rs11528010 Chr.10: 86876022-86876022 on Build GRCh38
rs749440726 Chr.10: 86875973-86875973 on Build GRCh38
rs1054345083 Chr.10: 86875936-86875936 on Build GRCh38
rs552863247 Chr.10: 86875983-86875983 on Build GRCh38
rs191498701 Chr.10: 86876110-86876110 on Build GRCh38
rs769464549 Chr.10: 86876115-86876115 on Build GRCh38
rs747777878 Chr.10: 86876111-86876111 on Build GRCh38
rs535411352 Chr.10: 86876051-86876051 on Build GRCh38
rs768904757 Chr.10: 86876101-86876101 on Build GRCh38
rs863224719 Chr.10: 86876040-86876040 on Build GRCh38
rs751420248 Chr.10: 86876045-86876045 on Build GRCh38
rs762926643 Chr.10: 86876037-86876037 on Build GRCh38
rs546681014 Chr.10: 86876011-86876011 on Build GRCh38
rs772729006 Chr.10: 86876030-86876030 on Build GRCh38
rs375966139 Chr.10: 86875965-86875965 on Build GRCh38
rs762442594 Chr.10: 86876118-86876118 on Build GRCh38
rs778886055 Chr.10: 86876067-86876067 on Build GRCh38

View Full Product Details