Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62415064-62415307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP8NL
Entrez Gene ID
140893
Gene Name
RBBP8 N-terminal like
Gene Aliases
C20orf151
UniGene
Hs.180374
Gene Chromosome Location
Chr.20: 62410237-62427573 on Build GRCh38
Transcripts
NM_080833.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3721056 c.708A>G p.R236R Chr.20: 62415207-62415207 on Build GRCh38
1713765 c.668C>T p.A223V Chr.20: 62415247-62415247 on Build GRCh38
7232488 c.794+7G>A p.? Chr.20: 62415114-62415114 on Build GRCh38
4648416 c.753C>T p.S251S Chr.20: 62415162-62415162 on Build GRCh38
4646023 c.725C>T p.T242M Chr.20: 62415190-62415190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937449034 Chr.20: 62415266-62415266 on Build GRCh38
rs62196317 Chr.20: 62415106-62415106 on Build GRCh38
rs376884862 Chr.20: 62415239-62415239 on Build GRCh38
rs369153183 Chr.20: 62415140-62415140 on Build GRCh38
rs764012856 Chr.20: 62415252-62415252 on Build GRCh38
rs773841918 Chr.20: 62415136-62415136 on Build GRCh38
rs770795292 Chr.20: 62415277-62415277 on Build GRCh38
rs764066164 Chr.20: 62415154-62415154 on Build GRCh38
rs760499152 Chr.20: 62415251-62415251 on Build GRCh38
rs759285229 Chr.20: 62415137-62415137 on Build GRCh38
rs762316630 Chr.20: 62415121-62415121 on Build GRCh38
rs940586083 Chr.20: 62415189-62415189 on Build GRCh38
rs878888099 Chr.20: 62415205-62415205 on Build GRCh38
rs767409193 Chr.20: 62415139-62415139 on Build GRCh38
rs36042460 Chr.20: 62415204-62415204 on Build GRCh38
rs781592550 Chr.20: 62415105-62415105 on Build GRCh38
rs551678730 Chr.20: 62415174-62415174 on Build GRCh38
rs768880033 Chr.20: 62415098-62415098 on Build GRCh38
rs373953870 Chr.20: 62415223-62415223 on Build GRCh38
rs372018613 Chr.20: 62415200-62415200 on Build GRCh38
rs770503692 Chr.20: 62415129-62415129 on Build GRCh38
rs6121576 Chr.20: 62415112-62415112 on Build GRCh38
rs6121577 Chr.20: 62415235-62415235 on Build GRCh38
rs766362259 Chr.20: 62415278-62415278 on Build GRCh38
rs751590926 Chr.20: 62415288-62415288 on Build GRCh38
rs35934795 Chr.20: 62415173-62415173 on Build GRCh38
rs571629533 Chr.20: 62415212-62415212 on Build GRCh38
rs1044922932 Chr.20: 62415214-62415214 on Build GRCh38
rs911079 Chr.20: 62415114-62415114 on Build GRCh38
rs550183119 Chr.20: 62415208-62415208 on Build GRCh38
rs560779290 Chr.20: 62415175-62415175 on Build GRCh38
rs368889241 Chr.20: 62415245-62415245 on Build GRCh38
rs751048977 Chr.20: 62415106-62415107 on Build GRCh38
rs200432639 Chr.20: 62415201-62415201 on Build GRCh38
rs1029138852 Chr.20: 62415115-62415115 on Build GRCh38
rs914944496 Chr.20: 62415184-62415184 on Build GRCh38
rs927670597 Chr.20: 62415147-62415147 on Build GRCh38
rs527948115 Chr.20: 62415190-62415190 on Build GRCh38
rs911565599 Chr.20: 62415184-62415186 on Build GRCh38
rs981983060 Chr.20: 62415145-62415145 on Build GRCh38

View Full Product Details